Learn More
1,3-Dichloro-2-iodo-5-(trifluoromethyl)benzene, 97%, Thermo Scientific™
£9.20 - £253.00
Chemical Identifiers
CAS | 175205-56-8 |
---|---|
Molecular Formula | C7H2Cl2F3I |
Molecular Weight (g/mol) | 340.89 |
MDL Number | MFCD00084997 |
InChI Key | RLCCFAVBZGICHD-UHFFFAOYSA-N |
Synonym | 1,3-dichloro-2-iodo-5-trifluoromethyl benzene, 3,5-dichloro-4-iodobenzotrifluoride, 3,5-dichloro-4-iodobenztrifluoride, buttpark 96\50-28, benzene,1,3-dichloro-2-iodo-5-trifluoromethyl, 1,3-bis chloranyl-2-iodanyl-5-trifluoromethyl benzene, 3,5-dichloro-4-iodo-alpha,alpha,alpha-trifluorotoluene |
PubChem CID | 2736838 |
IUPAC Name | 1,3-dichloro-2-iodo-5-(trifluoromethyl)benzene |
SMILES | FC(F)(F)C1=CC(Cl)=C(I)C(Cl)=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10180792
|
Thermo Scientific Maybridge
SEW01636DA |
1 g |
£9.20
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10140842
|
Thermo Scientific Maybridge
SEW01636EA |
10 g |
£62.15
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10747495
|
Thermo Scientific Maybridge
SEW01636EE |
50 g |
£253.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
175205-56-8 | |
340.89 | |
RLCCFAVBZGICHD-UHFFFAOYSA-N | |
2736838 | |
FC(F)(F)C1=CC(Cl)=C(I)C(Cl)=C1 |
C7H2Cl2F3I | |
MFCD00084997 | |
1,3-dichloro-2-iodo-5-trifluoromethyl benzene, 3,5-dichloro-4-iodobenzotrifluoride, 3,5-dichloro-4-iodobenztrifluoride, buttpark 96\50-28, benzene,1,3-dichloro-2-iodo-5-trifluoromethyl, 1,3-bis chloranyl-2-iodanyl-5-trifluoromethyl benzene, 3,5-dichloro-4-iodo-alpha,alpha,alpha-trifluorotoluene | |
1,3-dichloro-2-iodo-5-(trifluoromethyl)benzene |
Specifications
175205-56-8 | |
97% | |
C7H2Cl2F3I | |
1,3-dichloro-2-iodo-5-trifluoromethyl benzene, 3,5-dichloro-4-iodobenzotrifluoride, 3,5-dichloro-4-iodobenztrifluoride, buttpark 96\50-28, benzene,1,3-dichloro-2-iodo-5-trifluoromethyl, 1,3-bis chloranyl-2-iodanyl-5-trifluoromethyl benzene, 3,5-dichloro-4-iodo-alpha,alpha,alpha-trifluorotoluene | |
FC(F)(F)C1=CC(Cl)=C(I)C(Cl)=C1 | |
340.89 | |
340.89 | |
1, 3-Dichloro-2-iodo-5-(trifluoromethyl)benzene |
97.0 | |
Amber Glass Bottle | |
MFCD00084997 | |
RLCCFAVBZGICHD-UHFFFAOYSA-N | |
1,3-dichloro-2-iodo-5-(trifluoromethyl)benzene | |
2736838 | |
97% |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.