missing translation for 'onlineSavingsMsg'
Learn More
1,3-Dichloro-5,5-dimethylhydantoin, 98%, Thermo Scientific Chemicals Product Code.: 11410653

1,3-Dichloro-5,5-dimethylhydantoin, 98%, Thermo Scientific Chemicals

Product Code. 11410653
100 g
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy

Product Code. 11410653

Brand: Thermo Scientific Alfa Aesar A12718.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Dantoin(R) DCDMH is an industrial bleaching agent based on dimethyl hydantoin. This product finds application wherever solid bleach is desired. It is used as a mild chlorinating agent in organic synthesis.It is also used as a sanitizer in swimming pools, laundries restaurants, cutting oils and pharmaceutical industry as a general cleaner. Other uses include it is been used as a preventer and catalyst in polymer industry, used to emboss or texturize resinous sheet preparations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dantoin(R) DCDMH is an industrial bleaching agent based on dimethyl hydantoin. This product finds application wherever solid bleach is desired. It is used as a mild chlorinating agent in organic synthesis.It is also used as a sanitizer in swimming pools, laundries restaurants, cutting oils and pharmaceutical industry as a general cleaner. Other uses include it is been used as a preventer and catalyst in polymer industry, used to emboss or texturize resinous sheet preparations.

Solubility
Soluble in water 0.21 g/100 mL (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with reducing agents, acids, strong bases, organic materials, powdered metals. Moisture, light, air sensitive. Stable, but a strong oxidizer.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118-52-5
Molecular Formula C5H6Cl2N2O2
Molecular Weight (g/mol) 197.015
MDL Number MFCD00003190
InChI Key KEQGZUUPPQEDPF-UHFFFAOYSA-N
Synonym 1,3-dichloro-5,5-dimethylhydantoin, dantoin, dactin, daktin, dichlorantin, omchlor, halane, hydan, hydantoin, dichlorodimethyl, hydan antisepticShow MoreShow Less
PubChem CID 8360
IUPAC Name 1,3-dichloro-5,5-dimethylimidazolidine-2,4-dione
SMILES CC1(C(=O)N(C(=O)N1Cl)Cl)C

Specifications

Melting Point 130°C to 134°C
Density 1.5
Flash Point 174°C (345°F)
Quantity 100 g
UN Number UN1479
Beilstein 146013
Sensitivity Moisture sensitive
Merck Index 14,3065
Solubility Information Soluble in water 0.21g/100 mL (25°C).
Formula Weight 197.02
Percent Purity 98%
Chemical Name or Material 1,3-Dichloro-5,5-dimethylhydantoin
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10219498 Certificate TypeCertificate of Analysis Date13/01/2023 Catalog NumberA12718.22,A12718.36,A12718.0E
Lot Number10244562 Certificate TypeCertificate of Analysis Date28/11/2022 Catalog NumberA12718.22,A12718.36,A12718.0E
Lot Number10244437 Certificate TypeCertificate of Analysis Date03/11/2022 Catalog NumberA12718.22,A12718.36,A12718.0E
Lot NumberJ6336A Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12718.22,A12718.36,A12718.0E
Lot Number10195853 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12718.22,A12718.36,A12718.0E
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1,3-Dichloro-5,5-dimethylhydantoin
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 1,3-Dichloro-5,5-dimethylhydantoin

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,3-Dichloro-5,5-dimethylhydantoin, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.