missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichlorobenzene, 98%, Thermo Scientific Chemicals

£12.73 - £109.55

Chemical Identifiers

CAS 541-73-1
Molecular Formula C6H4Cl2
Molecular Weight (g/mol) 147
MDL Number MFCD00000573
InChI Key ZPQOPVIELGIULI-UHFFFAOYSA-N
Synonym m-dichlorobenzene, m-phenylene dichloride, m-dichlorobenzol, meta-dichlorobenzene, benzene, 1,3-dichloro, metadichlorobenzene, benzene, m-dichloro, m-phenylenedichloride, m-dcb, 3-dichlorobenzene
PubChem CID 10943
ChEBI CHEBI:36693
IUPAC Name 1,3-dichlorobenzene
SMILES C1=CC(=CC(=C1)Cl)Cl
View More Specs

Products 5
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10426990
View Documents Promotion Details
Thermo Scientific Acros
151180050
5 mL Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10635972
View Documents Promotion Details
Thermo Scientific Acros
151180250
25 mL Glass bottle
£16.50
25mL
Please to purchase this item. Need a web account? Register with us today!
10215530
View Documents Promotion Details
Thermo Scientific Acros
151182500
250 mL Glass bottle
£38.89
250mL
Please to purchase this item. Need a web account? Register with us today!
10377800
View Documents Promotion Details
Thermo Scientific Acros
151185000
500 mL Glass bottle
£61.51
500mL
Please to purchase this item. Need a web account? Register with us today!
10011810
View Documents Promotion Details
Thermo Scientific Acros
151180010
1 L Glass bottle
£109.55
1 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

541-73-1
147
ZPQOPVIELGIULI-UHFFFAOYSA-N
10943
1,3-dichlorobenzene
C6H4Cl2
MFCD00000573
m-dichlorobenzene, m-phenylene dichloride, m-dichlorobenzol, meta-dichlorobenzene, benzene, 1,3-dichloro, metadichlorobenzene, benzene, m-dichloro, m-phenylenedichloride, m-dcb, 3-dichlorobenzene
CHEBI:36693
C1=CC(=CC(=C1)Cl)Cl

Specifications

541-73-1
100.0
Colorless to Yellow
172°C to 173°C
Authentic
C6H4Cl2
1.5450 to 1.547
05, 202
1.28
Solubility in water: 0.0123g/100mL water (25°C). Other solubilities: soluble in organic solvents such as alcohol,,benzene,ether
C1=CC(=CC(=C1)Cl)Cl
147
CHEBI:36693
147
Liquid
97.5
-24°C
1.2800g/mL
67°C
97.5% min. (GC)
C6H4Cl2
MFCD00000573
15, 3064
m-dichlorobenzene, m-phenylene dichloride, m-dichlorobenzol, meta-dichlorobenzene, benzene, 1,3-dichloro, metadichlorobenzene, benzene, m-dichloro, m-phenylenedichloride, m-dcb, 3-dichlorobenzene
ZPQOPVIELGIULI-UHFFFAOYSA-N
1,3-dichlorobenzene
10943
1.045 mPa.s (23°C)
98%
1, 3-Dichlorobenzene, 98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic to aquatic life with long lasting effects.
Harmful if swallowed.

GHS P Statement:
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 208-792-1

missing translation for 'rtecsNumber' : CZ4499000

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only