Learn More
1,3-Difluoro-2-nitrobenzene, 98%, Thermo Scientific Chemicals
£16.68 - £214.00
Chemical Identifiers
CAS | 19064-24-5 |
---|---|
Molecular Formula | C6H3F2NO2 |
Molecular Weight (g/mol) | 159.092 |
MDL Number | MFCD00192035 |
InChI Key | SSNCMIDZGFCTST-UHFFFAOYSA-N |
Synonym | 2,6-difluoronitrobenzene, 1,3-difluoro-2-nitro-benzene, benzene, 1,3-difluoro-2-nitro, 2,6-difluoro nitrobenzene, pubchem4403, 1,3-difluoronitrobenzene, acmc-1buv1, 2,6-difluoro-nitrobenzene, 2,6-difluoro nitro benzene, 1,3-difluoro-2-nitrobenzen |
PubChem CID | 87922 |
IUPAC Name | 1,3-difluoro-2-nitrobenzene |
SMILES | C1=CC(=C(C(=C1)F)[N+](=O)[O-])F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15432018
|
Thermo Scientific Alfa Aesar
H55694.MD |
250 mg | |||||||
15412018
|
Thermo Scientific Alfa Aesar
H55694.03 |
1 g |
£54.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15422018
|
Thermo Scientific Alfa Aesar
H55694.06 |
5 g |
£214.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1,3-Difluoro-2-nitrobenzene, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,3-Difluoro-2-nitrobenzene, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, bases.
Chemical Identifiers
19064-24-5 | |
159.092 | |
SSNCMIDZGFCTST-UHFFFAOYSA-N | |
87922 | |
C1=CC(=C(C(=C1)F)[N+](=O)[O-])F |
C6H3F2NO2 | |
MFCD00192035 | |
2,6-difluoronitrobenzene, 1,3-difluoro-2-nitro-benzene, benzene, 1,3-difluoro-2-nitro, 2,6-difluoro nitrobenzene, pubchem4403, 1,3-difluoronitrobenzene, acmc-1buv1, 2,6-difluoro-nitrobenzene, 2,6-difluoro nitro benzene, 1,3-difluoro-2-nitrobenzen | |
1,3-difluoro-2-nitrobenzene |
Specifications
19064-24-5 | |
1.503 | |
88°C (190°F) | |
1.494 | |
2,6-difluoronitrobenzene, 1,3-difluoro-2-nitro-benzene, benzene, 1,3-difluoro-2-nitro, 2,6-difluoro nitrobenzene, pubchem4403, 1,3-difluoronitrobenzene, acmc-1buv1, 2,6-difluoro-nitrobenzene, 2,6-difluoro nitro benzene, 1,3-difluoro-2-nitrobenzen | |
SSNCMIDZGFCTST-UHFFFAOYSA-N | |
1,3-difluoro-2-nitrobenzene | |
87922 | |
98% |
0°C | |
91°C to 92°C (11 mmHg) | |
C6H3F2NO2 | |
MFCD00192035 | |
Slightly soluble in water. | |
C1=CC(=C(C(=C1)F)[N+](=O)[O-])F | |
159.092 | |
159.09 | |
1,3-Difluoro-2-nitrobenzene |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
H227-H302+H312+H332-H315-H319-H335
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III
missing translation for 'einecsNumber' : 242-793-8
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only