Learn More
1,3-Dimethyl-1H-thieno[2,3-c]pyrazole-5-carbonyl chloride, 97%, Thermo Scientific™
£73.60 - £117.00
Chemical Identifiers
CAS | 388088-75-3 |
---|---|
Molecular Formula | C8H7ClN2OS |
Molecular Weight (g/mol) | 214.67 |
MDL Number | MFCD03086153 |
InChI Key | BQOTYHLFKDBAGL-UHFFFAOYSA-N |
Synonym | 1,3-dimethyl-1h-thieno 2,3-c pyrazole-5-carbonyl chloride, 1,3-dimethylthieno 2,3-c pyrazole-5-carbonyl chloride, 1,3-dimethyl-1h-thieno 2,3-c pyrazole-5-carbonylchloride, 1,3-dimethylthiopheno 3,2-d pyrazole-5-carbonyl chloride, 1h-thieno 2,3-c pyrazole-5-carbonylchloride, 1,3-dimethyl |
PubChem CID | 2776542 |
IUPAC Name | 1,3-dimethylthieno[2,3-c]pyrazole-5-carbonyl chloride |
SMILES | CN1N=C(C)C2=C1SC(=C2)C(Cl)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10416852
|
Thermo Scientific Maybridge
CC22902CB |
250 mg | Amber Glass Bottle |
£73.60
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10522862
|
Thermo Scientific Maybridge
CC22902DA |
1 g | Amber Glass Bottle |
£117.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
388088-75-3 | |
214.67 | |
BQOTYHLFKDBAGL-UHFFFAOYSA-N | |
2776542 | |
CN1N=C(C)C2=C1SC(=C2)C(Cl)=O |
C8H7ClN2OS | |
MFCD03086153 | |
1,3-dimethyl-1h-thieno 2,3-c pyrazole-5-carbonyl chloride, 1,3-dimethylthieno 2,3-c pyrazole-5-carbonyl chloride, 1,3-dimethyl-1h-thieno 2,3-c pyrazole-5-carbonylchloride, 1,3-dimethylthiopheno 3,2-d pyrazole-5-carbonyl chloride, 1h-thieno 2,3-c pyrazole-5-carbonylchloride, 1,3-dimethyl | |
1,3-dimethylthieno[2,3-c]pyrazole-5-carbonyl chloride |
Specifications
388088-75-3 | |
100.0 | |
C8H7ClN2OS | |
1,3-dimethyl-1h-thieno 2,3-c pyrazole-5-carbonyl chloride, 1,3-dimethylthieno 2,3-c pyrazole-5-carbonyl chloride, 1,3-dimethyl-1h-thieno 2,3-c pyrazole-5-carbonylchloride, 1,3-dimethylthiopheno 3,2-d pyrazole-5-carbonyl chloride, 1h-thieno 2,3-c pyrazole-5-carbonylchloride, 1,3-dimethyl | |
CN1N=C(C)C2=C1SC(=C2)C(Cl)=O | |
214.67 | |
214.68 | |
1, 3-Dimethyl-1H-thieno[2, 3-c]pyrazole-5-carbonyl chloride |
96.5 | |
97% | |
MFCD03086153 | |
BQOTYHLFKDBAGL-UHFFFAOYSA-N | |
1,3-dimethylthieno[2,3-c]pyrazole-5-carbonyl chloride | |
2776542 | |
97% |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.