missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Thiazol-2-ylmethanol, 97+%, Thermo Scientific™
£100.00 - £691.00
Chemical Identifiers
CAS | 14542-12-2 |
---|---|
Molecular Formula | C4H5NOS |
Molecular Weight (g/mol) | 115.15 |
MDL Number | MFCD06200855 |
InChI Key | JNHDLNXNYPLBMJ-UHFFFAOYSA-N |
Synonym | thiazol-2-ylmethanol, 2-thiazolemethanol, thiazol-2-yl-methanol, 2-hydroxymethylthiazole, 2-hydroxymethyl thiazole, 2-hydroxymehtylthiazole, 1,3-thiazol-2-yl methanol, thiazol-2-yl methanol, thiazole-2-methanol, 2-thiazolylmethanol |
PubChem CID | 2795213 |
SMILES | OCC1=NC=CS1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10636463
|
Thermo Scientific Maybridge
CC09509DA |
1 g | Amber Glass Bottle |
£100.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10623595
|
Thermo Scientific Maybridge
CC09509DE |
5 g | Amber Glass Bottle |
£370.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10376964
|
Thermo Scientific Maybridge
CC09509EA |
10 g | Amber Glass Bottle |
£691.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
14542-12-2 | |
115.15 | |
JNHDLNXNYPLBMJ-UHFFFAOYSA-N | |
2795213 |
C4H5NOS | |
MFCD06200855 | |
thiazol-2-ylmethanol, 2-thiazolemethanol, thiazol-2-yl-methanol, 2-hydroxymethylthiazole, 2-hydroxymethyl thiazole, 2-hydroxymehtylthiazole, 1,3-thiazol-2-yl methanol, thiazol-2-yl methanol, thiazole-2-methanol, 2-thiazolylmethanol | |
OCC1=NC=CS1 |
Specifications
14542-12-2 | |
C4H5NOS | |
thiazol-2-ylmethanol, 2-thiazolemethanol, thiazol-2-yl-methanol, 2-hydroxymethylthiazole, 2-hydroxymethyl thiazole, 2-hydroxymehtylthiazole, 1,3-thiazol-2-yl methanol, thiazol-2-yl methanol, thiazole-2-methanol, 2-thiazolylmethanol | |
OCC1=NC=CS1 | |
115.15 | |
115.16 | |
1, 3-Thiazol-2-ylmethanol |
97% | |
MFCD06200855 | |
JNHDLNXNYPLBMJ-UHFFFAOYSA-N | |
1,3-thiazol-2-ylmethanol | |
2795213 | |
97% |
Safety and Handling
Warning
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
1,3-Thiazol-2-ylmethanol, 97+%, Thermo Scientific™