missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-[4-(4-Morpholinyl)phenyl]guanidine, 98%, Thermo Scientific Chemicals
£48.00 - £492.00
Chemical Identifiers
CAS | 247234-41-9 |
---|---|
Molecular Formula | C11H16N4O |
Molecular Weight (g/mol) | 220.28 |
MDL Number | MFCD11986905 |
InChI Key | ZVZJREQBRCRGLM-UHFFFAOYSA-N |
Synonym | 1-4-morpholinophenyl guanidine, n-4-morpholin-4-ylphenyl guanidine, 4-morpholinophenylguanidine, 4-morpholino phenylguanidine, n-4-morpholin-4-yl-phenyl-guanidine, n-4-morpholin-4-yl phenyl guanidine, 1-4-4-morpholinyl phenyl guanidine, 1-4-morpholin-4-yl phenyl guanidine, 2-4-morpholin-4-yl phenyl guanidine |
PubChem CID | 10176830 |
IUPAC Name | 2-(4-morpholin-4-ylphenyl)guanidine |
SMILES | NC(N)=NC1=CC=C(C=C1)N1CCOCC1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15436577
|
Thermo Scientific Alfa Aesar
H33726.03 |
1 g |
£48.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15446577
|
Thermo Scientific Alfa Aesar
H33726.06 |
5 g |
£153.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15456577
|
Thermo Scientific Alfa Aesar
H33726.14 |
25 g |
£492.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
247234-41-9 | |
220.28 | |
ZVZJREQBRCRGLM-UHFFFAOYSA-N | |
10176830 | |
NC(N)=NC1=CC=C(C=C1)N1CCOCC1 |
C11H16N4O | |
MFCD11986905 | |
1-4-morpholinophenyl guanidine, n-4-morpholin-4-ylphenyl guanidine, 4-morpholinophenylguanidine, 4-morpholino phenylguanidine, n-4-morpholin-4-yl-phenyl-guanidine, n-4-morpholin-4-yl phenyl guanidine, 1-4-4-morpholinyl phenyl guanidine, 1-4-morpholin-4-yl phenyl guanidine, 2-4-morpholin-4-yl phenyl guanidine | |
2-(4-morpholin-4-ylphenyl)guanidine |
Specifications
247234-41-9 | |
1.309 | |
MFCD11986905 | |
ZVZJREQBRCRGLM-UHFFFAOYSA-N | |
2-(4-morpholin-4-ylphenyl)guanidine | |
10176830 | |
98% |
∼220°C (decomposition) | |
C11H16N4O | |
1-4-morpholinophenyl guanidine, n-4-morpholin-4-ylphenyl guanidine, 4-morpholinophenylguanidine, 4-morpholino phenylguanidine, n-4-morpholin-4-yl-phenyl-guanidine, n-4-morpholin-4-yl phenyl guanidine, 1-4-4-morpholinyl phenyl guanidine, 1-4-morpholin-4-yl phenyl guanidine, 2-4-morpholin-4-yl phenyl guanidine | |
NC(N)=NC1=CC=C(C=C1)N1CCOCC1 | |
220.28 | |
220.27 | |
1-[4-(4-Morpholinyl)phenyl]guanidine |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only