missing translation for 'onlineSavingsMsg'
Learn More

1,4,7,10-Tetraazacyclotridecane, Thermo Scientific Chemicals

£50.10 - £145.00

Chemical Identifiers

CAS 295-14-7
Molecular Formula C9H22N4
Molecular Weight (g/mol) 186.303
MDL Number MFCD06410986
InChI Key LADZJJOUGVGJHM-UHFFFAOYSA-N
Synonym 1,4,7,10-tetraazacyclotridecane, homocyclen, acmc-20ap3n, 13 anen4
PubChem CID 11478723
IUPAC Name 1,4,7,10-tetrazacyclotridecane
SMILES C1CNCCNCCNCCNC1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15473517
View Documents
Thermo Scientific Alfa Aesar
H26671.MD
250 mg
£50.10
250mg
Please to purchase this item. Need a web account? Register with us today!
11372589
View Documents
Thermo Scientific Alfa Aesar
H26671.03
1 g
£145.00
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

1,4,7,10-Tetraazacyclotridecane is used to find the binding constants, and for building block elements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4,7,10-Tetraazacyclotridecane is used to find the binding constants, and for building block elements.

Solubility
Soluble in water.

Notes
Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

295-14-7
186.303
LADZJJOUGVGJHM-UHFFFAOYSA-N
11478723
C1CNCCNCCNCCNC1
C9H22N4
MFCD06410986
1,4,7,10-tetraazacyclotridecane, homocyclen, acmc-20ap3n, 13 anen4
1,4,7,10-tetrazacyclotridecane

Specifications

295-14-7
MFCD06410986
1,4,7,10-tetraazacyclotridecane, homocyclen, acmc-20ap3n, 13 anen4
LADZJJOUGVGJHM-UHFFFAOYSA-N
1,4,7,10-tetrazacyclotridecane
11478723
1,4,7,10-Tetraazacyclotridecane
C9H22N4
Hygroscopic
Soluble in water.
C1CNCCNCCNCCNC1
186.303
186.3
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only