missing translation for 'onlineSavingsMsg'
Learn More
1,4-Dioxane, 99+%, ACS reagent, stabilized, meets the requirements of Reag.Ph.Eur., Thermo Scientific Chemicals Product Code.: 15189273

1,4-Dioxane, 99+%, ACS reagent, stabilized, meets the requirements of Reag.Ph.Eur., Thermo Scientific Chemicals

Product Code. 15189273
10 L, Plastic Drum
Click to view available options
Quantity:
500 mL
1 L
2.5 L
10 L
Packaging:
EcoSafPak™ Glass Bottle
Glass Bottle
Plastic Drum
This item is not returnable. View return policy

Product Code. 15189273

Brand: Thermo Scientific Acros 408820100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-91-1
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00006571
InChI Key RYHBNJHYFVUHQT-UHFFFAOYSA-N
Synonym dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxinShow MoreShow Less
PubChem CID 31275
ChEBI CHEBI:47032
IUPAC Name 1,4-dioxane
SMILES C1COCCO1

Specifications

Melting Point 12.0°C
CAS Min % 99.0
CAS Max % 100.0
Density 1.0340g/mL
Boiling Point 101.0°C
Quantity 10 L
Flash Point 12°C
Packaging Plastic Drum
Physical Form Liquid
Refractive Index 1.4205 to 1.4220
Assay Percent Range 99+%
Residue after Evaporation 0.005% max.
Specific Gravity 1.034
Titratable Acid 0.0016 meq/g max.
Viscosity 1.32 mPa.s (20°C)
Beilstein 19, 3
Water 0.05% max. (Coulometric)
Fieser 01,333
Merck Index 15, 3328
Formula Weight 88.11
Percent Purity ≥99%
Grade ACS Reagent
Chemical Name or Material 1, 4-Dioxane
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0461760 Certificate TypeCertificate of Analysis Date25/04/2024 Catalog Number408820100,408825000,408820010,408820025
Lot NumberA0459973 Certificate TypeCertificate of Analysis Date06/02/2024 Catalog Number408820100,408825000,408820010,408820025
Lot NumberB0541156A Certificate TypeCertificate of Analysis Date14/11/2023 Catalog Number408820100,408825000,408820010,408820025
Lot NumberA0445678 Certificate TypeCertificate of Analysis Date01/09/2023 Catalog Number408820100,408825000,408820010,408820025
Lot NumberA0453881 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number408820100,408825000,408820010,408820025

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,4-Dioxane, 99+%, ACS reagent, stabilized, meets the requirements of Reag.Ph.Eur., Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.