missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Dithiothreitol, Ultrapure Bioreagent, J.T. Baker™
High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography
£749.00 - £5245.00
Chemical Identifiers
CAS | 3483-12-3 |
---|---|
Molecular Formula | C4H10O2S2 |
Molecular Weight (g/mol) | 154.24 |
InChI Key | VHJLVAABSRFDPM-IMJSIDKUSA-N |
Synonym | dithiothreitol, dl-1,4-dithiothreitol, dl-dithiothreitol, 1,4-dithio-dl-threitol, d-1,4-dithiothreitol, d-dtt, 2s,3s-1,4-dimercaptobutane-2,3-diol, threo-1,4-dimercapto-2,3-butanediol, 1,4-dithiothreitol, dtt |
PubChem CID | 446094 |
ChEBI | CHEBI:42170 |
IUPAC Name | (2S,3S)-1,4-bis(sulfanyl)butane-2,3-diol |
SMILES | C(C(C(CS)O)O)S |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15548774
|
Avantor J.T.Baker
F780-01 |
5 g | POLYSTORMOR | ||||||
15558774
|
Avantor J.T.Baker
F780-02 |
25 g | POLYSTORMOR | ||||||
15568774
|
Avantor J.T.Baker
F780-03 |
100 g | Poly Bottle |
£5245.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- For Laboratory,Research,or Manufacturing Use
Chemical Identifiers
3483-12-3 | |
154.24 | |
dithiothreitol, dl-1,4-dithiothreitol, dl-dithiothreitol, 1,4-dithio-dl-threitol, d-1,4-dithiothreitol, d-dtt, 2s,3s-1,4-dimercaptobutane-2,3-diol, threo-1,4-dimercapto-2,3-butanediol, 1,4-dithiothreitol, dtt | |
CHEBI:42170 | |
C(C(C(CS)O)O)S |
C4H10O2S2 | |
VHJLVAABSRFDPM-IMJSIDKUSA-N | |
446094 | |
(2S,3S)-1,4-bis(sulfanyl)butane-2,3-diol |
Specifications
3483-12-3 | |
C4H10O2S2 | |
dithiothreitol, dl-1,4-dithiothreitol, dl-dithiothreitol, 1,4-dithio-dl-threitol, d-1,4-dithiothreitol, d-dtt, 2s,3s-1,4-dimercaptobutane-2,3-diol, threo-1,4-dimercapto-2,3-butanediol, 1,4-dithiothreitol, dtt | |
C(C(C(CS)O)O)S | |
154.24 | |
CHEBI:42170 | |
Ultrapure Bioreagent |
109.00°C | |
1L = 1.00kg | |
VHJLVAABSRFDPM-IMJSIDKUSA-N | |
(2S,3S)-1,4-bis(sulfanyl)butane-2,3-diol | |
446094 | |
154.25 | |
1,4-Dithiothreitol |