missing translation for 'onlineSavingsMsg'
Learn More
1,4-Hexadiene, cis + trans, 99%, Thermo Scientific Chemicals Product Code.: 11480541

1,4-Hexadiene, cis + trans, 99%, Thermo Scientific Chemicals

Product Code. 11480541
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11480541

Brand: Thermo Scientific Alfa Aesar B20060.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 592-45-0
Molecular Formula C6H10
Molecular Weight (g/mol) 82.146
MDL Number MFCD00009298
InChI Key PRBHEGAFLDMLAL-GQCTYLIASA-N
Synonym 1,4-hexadiene, trans-1,4-hexadiene, 1,4-hexadiene, 4e, 1,4-hexadiene,c&t, allylpropenyl, 1,4-hexadiene cis-trans, 1,4-hexa-1,4-diene, 1-allylpropene, 1,4-hexadiene,cis-trans, 1,4-trans-hexadiene
PubChem CID 5365552
IUPAC Name (4E)-hexa-1,4-diene
SMILES CC=CCC=C

Specifications

Density 0.71
Boiling Point 64°C to 66°C
Flash Point −26°C (−15°F)
Odor Characteristic
Refractive Index 1.411
Quantity 5 g
UN Number UN2458
Beilstein 1731889
Formula Weight 82.15
Percent Purity 99%
Chemical Name or Material 1,4-Hexadiene, cis + trans
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10247429 Certificate TypeCertificate of Analysis Date25/07/2023 Catalog NumberB20060.14,B20060.06
Lot Number10241066 Certificate TypeCertificate of Analysis Date09/06/2022 Catalog NumberB20060.14,B20060.06
Lot Number10177936 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB20060.14,B20060.06
Lot NumberG9700A Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB20060.14,B20060.06
Lot NumberG1923B Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB20060.14,B20060.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,4-Hexadiene, cis + trans, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.