missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gilac™ 1/4 Turn Valve for 310 L and 500 L Double-wall Containers
1/4 Turn Valve for 310 L and 500 L Double-wall Containers ensures high velocity drainage.
Brand: Gilac™ G119441
This item is not returnable.
View return policy
Description
- 1/4 turn valve for 310 L or 500 L double-wall containers.
- Valve that ensures high velocity drainage.
- Food contact material.
- Drainage tube may be installed.
Specifications
Spigot | |
310 L and 500 L Double-wall Containers |
Product Suggestions
Customers who viewed this item also viewed.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Gilac™ 1/4 Turn Valve for 310 L and 500 L Double-wall Containers > 1/4 Turn Valve for 310 L and 500 L Double-wall Containers
Spot an opportunity for improvement?Share a Content Correction