missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ LabMat™ Bench Liners
Simplify cleanup and safeguard work surfaces from stains, spills and wear. Bel Art™ SP Scienceware™ LabMat™ Bench Liners can be rolled up and discard after contamination.
Brand: Bel-Art™ F24675-0000
117.61 GBP valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Description
- Highly absorbent upper surface traps up to 420mL/m2 of water, dyes, corrosive chemicals and radioactive solutions
- Flexible polyethylene film backing is nonskid and prevents soak-through of most solutions
- Cut to size easily with scissors or the Scienceware™ Safety Cutter (Mfr. No. 21012-0000)
- Roll up and discard in proper waste container after contamination
- Select orange to identify use with biohazard materials
Specifications
Bench Protector | |
400 mL/m2 | |
15.2 m | |
508 mm | |
White | |
Single-use |
Polyethylene | |
50 ft. | |
20 in. | |
50 ft. x 20 in. (15.2m x 508mm) | |
One single roll, 50 ft. | |
Simplify cleanup and safeguard work surfaces from stains, spills and wear |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Bel-Art™ SP Scienceware™ LabMat™ Bench Liners > 50 ft. Roll, White
Spot an opportunity for improvement?Share a Content Correction