missing translation for 'onlineSavingsMsg'
Learn More

1,5-Diamino-4,8-dihydroxy-9,10-dihydroanthracene-9,10-dione, Tech., Thermo Scientific™

£23.00 - £171.00

Chemical Identifiers

CAS 145-49-3
Molecular Formula C14H10N2O4
Molecular Weight (g/mol) 270.244
MDL Number MFCD00013722
InChI Key HSYLKWSCFRLSKB-UHFFFAOYSA-N
Synonym 1,5-diamino-4,8-dihydroxyanthraquinone, diaminoanthrarufin, 1,5-diaminoanthrarufin, 4,8-diaminoanthrarufin, 1,5-dihydroxy-4,8-diaminoanthraquinone, anthrarufin, 4,8-diamino, unii-q7yvg56q18, ccris 5599, 4,8-diamino-1,5-dihydroxyanthraquinone, 1,5-diamino-4,8-dihydroxy-anthracene-9,10-dione
PubChem CID 67351
IUPAC Name 1,5-diamino-4,8-dihydroxyanthracene-9,10-dione
SMILES C1=CC(=C2C(=C1N)C(=O)C3=C(C=CC(=C3C2=O)N)O)O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10213662
View Documents
Thermo Scientific Maybridge
SB01090EA
10 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10131082
View Documents
Thermo Scientific Maybridge
SB01090EE
50 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10554695
View Documents
Thermo Scientific Maybridge
SB01090FA
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Chemical Identifiers

145-49-3
270.244
HSYLKWSCFRLSKB-UHFFFAOYSA-N
67351
C1=CC(=C2C(=C1N)C(=O)C3=C(C=CC(=C3C2=O)N)O)O
C14H10N2O4
MFCD00013722
1,5-diamino-4,8-dihydroxyanthraquinone, diaminoanthrarufin, 1,5-diaminoanthrarufin, 4,8-diaminoanthrarufin, 1,5-dihydroxy-4,8-diaminoanthraquinone, anthrarufin, 4,8-diamino, unii-q7yvg56q18, ccris 5599, 4,8-diamino-1,5-dihydroxyanthraquinone, 1,5-diamino-4,8-dihydroxy-anthracene-9,10-dione
1,5-diamino-4,8-dihydroxyanthracene-9,10-dione

Specifications

145-49-3
C14H10N2O4
1,5-diamino-4,8-dihydroxyanthraquinone, diaminoanthrarufin, 1,5-diaminoanthrarufin, 4,8-diaminoanthrarufin, 1,5-dihydroxy-4,8-diaminoanthraquinone, anthrarufin, 4,8-diamino, unii-q7yvg56q18, ccris 5599, 4,8-diamino-1,5-dihydroxyanthraquinone, 1,5-diamino-4,8-dihydroxy-anthracene-9,10-dione
C1=CC(=C2C(=C1N)C(=O)C3=C(C=CC(=C3C2=O)N)O)O
270.244
270.24
Technical
Amber Glass Bottle
MFCD00013722
HSYLKWSCFRLSKB-UHFFFAOYSA-N
1,5-diamino-4,8-dihydroxyanthracene-9,10-dione
67351
Tech.
1,5-Diamino-4,8-dihydroxy-9,10-dihydroanthracene-9,10-dione
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.