missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One 1.5 mL Polypropylene Microcentrifuge Tubes

For single use only. Greiner Bio-One Cell 1.5mL Polypropylene Microcentrifuge Tubes are free of heavy metal.

Brand:  Greiner Bio-One 616274

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11729502

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

  • Material: Polypropylene
  • For single use only
  • Available in different colors
  • Free of detectable DNase/RNase, human DNA and pyrogens
  • Suitable for Eppendorf centrifuges and systems
Specifications

Specifications

With cap
Polypropylene
Non-sterile
Blue
1.5mL
Yes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers