Learn More
1,6-Diisocyanatohexane, 99+%, Thermo Scientific Chemicals
£24.70 - £186.00
Chemical Identifiers
CAS | 822-06-0 |
---|---|
Molecular Formula | C8H12N2O2 |
Molecular Weight (g/mol) | 168.20 |
MDL Number | MFCD00002047 |
InChI Key | RRAMGCGOFNQTLD-UHFFFAOYSA-N |
Synonym | hexamethylene diisocyanate, 1,6-hexamethylene diisocyanate, hmdi, hexane, 1,6-diisocyanato, hexane 1,6-diisocyanate, 1,6-hexylene diisocyanate, hexamethylene-1,6-diisocyanate, 1,6-hexanediol diisocyanate, hdi, isocyanic acid, hexamethylene ester |
PubChem CID | 13192 |
ChEBI | CHEBI:53578 |
IUPAC Name | 1,6-diisocyanatohexane |
SMILES | O=C=NCCCCCCN=C=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10336130
|
Thermo Scientific
115190250 |
25 g | Glass bottle |
£24.70
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10408260
|
Thermo Scientific Acros
115191000 |
100 g | Glass bottle |
£34.30
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10772623
|
Thermo Scientific Acros
115190010 |
1 kg | Glass bottle |
£186.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
822-06-0 | |
168.20 | |
RRAMGCGOFNQTLD-UHFFFAOYSA-N | |
13192 | |
1,6-diisocyanatohexane |
C8H12N2O2 | |
MFCD00002047 | |
hexamethylene diisocyanate, 1,6-hexamethylene diisocyanate, hmdi, hexane, 1,6-diisocyanato, hexane 1,6-diisocyanate, 1,6-hexylene diisocyanate, hexamethylene-1,6-diisocyanate, 1,6-hexanediol diisocyanate, hdi, isocyanic acid, hexamethylene ester | |
CHEBI:53578 | |
O=C=NCCCCCCN=C=O |
Specifications
822-06-0 | |
100.0 | |
Colorless to Yellow | |
255.0°C | |
Authentic | |
C8H12N2O2 | |
1.4510 to 1.4530 | |
04, II, 711 | |
hexamethylene diisocyanate, 1,6-hexamethylene diisocyanate, hmdi, hexane, 1,6-diisocyanato, hexane 1,6-diisocyanate, 1,6-hexylene diisocyanate, hexamethylene-1,6-diisocyanate, 1,6-hexanediol diisocyanate, hdi, isocyanic acid, hexamethylene ester | |
RRAMGCGOFNQTLD-UHFFFAOYSA-N | |
1,6-diisocyanatohexane | |
13192 | |
168.19 | |
Liquid |
99.0 | |
-67°C | |
1.0400g/mL | |
130°C | |
99% min. (GC) | |
OCN(CH2)6NCO | |
MFCD00002047 | |
1.04 | |
Solubility in water: decomposes. Other solubilities: soluble in benzene,acetone,toluene,,ethyl acetate,chlorobenzene,very soluble in ether | |
O=C=NCCCCCCN=C=O | |
168.20 | |
CHEBI:53578 | |
99+% | |
1, 6-Diisocyanatohexane, 99+% |
Safety and Handling
GHS H Statement
Fatal if inhaled.
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful
GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hai
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 212-485-8
missing translation for 'rtecsNumber' : MO1740000
missing translation for 'tsca' : TSCA
- 1,6-Diisocyanatohexane
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H331-Toxic if inhaled.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P311-Call a POISON CENTER/doctor/ .
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only