Learn More
1,6-Hexanediol diacrylate, 99% (reactive esters), stab. with 90ppm hydroquinone, Thermo Scientific Chemicals
£37.70 - £135.00
Chemical Identifiers
CAS | 13048-33-4 |
---|---|
Molecular Formula | C12H18O4 |
Molecular Weight (g/mol) | 226.27 |
MDL Number | MFCD00008631 |
InChI Key | FIHBHSQYSYVZQE-UHFFFAOYSA-N |
Synonym | 1,6-hexanediol diacrylate, hexamethylene diacrylate, kayarad hdda, hexane-1,6-diyl diacrylate, 1,6-hexamethylene diacrylate, hexamethylene glycol diacrylate, sartomer 238, hdoda, viscoat 230, hexamethylene acrylate |
PubChem CID | 25644 |
IUPAC Name | 6-prop-2-enoyloxyhexyl prop-2-enoate |
SMILES | C=CC(=O)OCCCCCCOC(=O)C=C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11349268
|
Thermo Scientific Alfa Aesar
043203.18 |
50 g |
£37.70
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11339268
|
Thermo Scientific Alfa Aesar
043203.30 |
250 g |
£135.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1,6-Hexanediol diacrylate is used as a functional monomer for polymers. It acts as a cross linking agent between the molecular chains of polymers. Further, it is used in adhesives, sealants, alkyd coatings, elastomers, photopolymers, and inks for improved adhesion, hardness, abrasion and heat resistance.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13048-33-4 | |
226.27 | |
FIHBHSQYSYVZQE-UHFFFAOYSA-N | |
25644 | |
C=CC(=O)OCCCCCCOC(=O)C=C |
C12H18O4 | |
MFCD00008631 | |
1,6-hexanediol diacrylate, hexamethylene diacrylate, kayarad hdda, hexane-1,6-diyl diacrylate, 1,6-hexamethylene diacrylate, hexamethylene glycol diacrylate, sartomer 238, hdoda, viscoat 230, hexamethylene acrylate | |
6-prop-2-enoyloxyhexyl prop-2-enoate |
Specifications
13048-33-4 | |
>110°C (>230°F) | |
MFCD00008631 | |
1,6-hexanediol diacrylate, hexamethylene diacrylate, kayarad hdda, hexane-1,6-diyl diacrylate, 1,6-hexamethylene diacrylate, hexamethylene glycol diacrylate, sartomer 238, hdoda, viscoat 230, hexamethylene acrylate | |
FIHBHSQYSYVZQE-UHFFFAOYSA-N | |
6-prop-2-enoyloxyhexyl prop-2-enoate | |
25644 | |
99% | |
(reactive esters) |
1.010 g/mL | |
C12H18O4 | |
Hygroscopic | |
Slightly miscible with water. | |
C=CC(=O)OCCCCCCOC(=O)C=C | |
226.27 | |
226.28 | |
Liquid | |
1,6-Hexanediol diacrylate, Stabilized with 90ppm hydroquinone |
Safety and Handling
GHS H Statement
H315-H319-H317
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
P261-P264b-P272-P280-P302+P352-P305+P351+P338-P333+P313-P362-P501c
H315-H317-H319
missing translation for 'einecsNumber' : 235-921-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only