missing translation for 'onlineSavingsMsg'
Learn More

1,8-Diazafluoren-9-one, Thermo Scientific Chemicals

£234.17 - £697.47

Chemical Identifiers

CAS 54078-29-4
Molecular Formula C11H6N2O
Molecular Weight (g/mol) 182.18
InChI Key FOSUVSBKUIWVKI-UHFFFAOYSA-N
Synonym 9h-cyclopenta 1,2-b:4,3-b' dipyridin-9-one, 1,8-diazafluoren-9-one, 1,8-diazafluoran-9-one, 9h-pyrido 3',2':3,4 cyclopenta 1,2-b pyridin-9-one, 6,10-diazatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2,4,6,10,12-hexaen-8-one, 1.8-diazafluoren-9-one, 9h-1,8-diazafluoren-9-one, fosuvsbkuiwvki-uhfffaoysa, pyridino 2',3'-1,5 cyclopenta 3,4-b pyridin-9-one, 6,10-diazatricyclo 7.4.0.0^ 2,7 trideca-1 9 ,2,4,6,10,12-hexaen-8-one
PubChem CID 725961
SMILES C1=CC2=C(C(=O)C3=C2C=CC=N3)N=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15730909
View Documents Promotion Details
Thermo Scientific Acros
461310010
1 g
£234.17
1g
Please to purchase this item. Need a web account? Register with us today!
15740909
View Documents Promotion Details
Thermo Scientific Acros
461310050
5 g
£697.47
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

54078-29-4
182.18
9h-cyclopenta 1,2-b:4,3-b' dipyridin-9-one, 1,8-diazafluoren-9-one, 1,8-diazafluoran-9-one, 9h-pyrido 3',2':3,4 cyclopenta 1,2-b pyridin-9-one, 6,10-diazatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2,4,6,10,12-hexaen-8-one, 1.8-diazafluoren-9-one, 9h-1,8-diazafluoren-9-one, fosuvsbkuiwvki-uhfffaoysa, pyridino 2',3'-1,5 cyclopenta 3,4-b pyridin-9-one, 6,10-diazatricyclo 7.4.0.0^ 2,7 trideca-1 9 ,2,4,6,10,12-hexaen-8-one
C1=CC2=C(C(=O)C3=C2C=CC=N3)N=C1
C11H6N2O
FOSUVSBKUIWVKI-UHFFFAOYSA-N
725961

Specifications

54078-29-4
C11H6N2O
FOSUVSBKUIWVKI-UHFFFAOYSA-N
182.18
182.18
229.0°C to 233.0°C
9h-cyclopenta 1,2-b:4,3-b' dipyridin-9-one, 1,8-diazafluoren-9-one, 1,8-diazafluoran-9-one, 9h-pyrido 3',2':3,4 cyclopenta 1,2-b pyridin-9-one, 6,10-diazatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2,4,6,10,12-hexaen-8-one, 1.8-diazafluoren-9-one, 9h-1,8-diazafluoren-9-one, fosuvsbkuiwvki-uhfffaoysa, pyridino 2',3'-1,5 cyclopenta 3,4-b pyridin-9-one, 6,10-diazatricyclo 7.4.0.0^ 2,7 trideca-1 9 ,2,4,6,10,12-hexaen-8-one
C1=CC2=C(C(=O)C3=C2C=CC=N3)N=C1
725961
1,8-Diazafluoren-9-one
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

Product Identifier
  • 1,8-Diazafluoren-9-one

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : 1,8-Diazafluoren-9-one

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only