missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Benzyl-3-pyrrolidinone, 98%, Thermo Scientific Chemicals
£48.30 - £620.00
Chemical Identifiers
CAS | 775-16-6 |
---|---|
Molecular Formula | C11H13NO |
Molecular Weight (g/mol) | 175.231 |
MDL Number | MFCD00005342 |
InChI Key | DHGMDHQNUNRMIN-UHFFFAOYSA-N |
Synonym | 1-benzyl-3-pyrrolidinone, n-benzyl-3-pyrrolidinone, 1-phenylmethyl-3-pyrrolidinone, 1-benzyl-3-pyrrolidone, 1-benzyl-pyrrolidin-3-one, 1-benzyl pyrrolidin-3-one, 3-pyrrolidinone, 1-phenylmethyl, benzyl-3-pyrrolidinone, pubchem10007, n-benzylpyrrolid-3-one |
PubChem CID | 69890 |
IUPAC Name | 1-benzylpyrrolidin-3-one |
SMILES | C1CN(CC1=O)CC2=CC=CC=C2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11424200
|
Thermo Scientific Alfa Aesar
A13426.03 |
1 g |
£48.30
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11434200
|
Thermo Scientific Alfa Aesar
A13426.06 |
5 g | |||||||
11485463
|
Thermo Scientific Alfa Aesar
A13426.14 |
25 g |
£620.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
775-16-6 | |
175.231 | |
DHGMDHQNUNRMIN-UHFFFAOYSA-N | |
69890 | |
C1CN(CC1=O)CC2=CC=CC=C2 |
C11H13NO | |
MFCD00005342 | |
1-benzyl-3-pyrrolidinone, n-benzyl-3-pyrrolidinone, 1-phenylmethyl-3-pyrrolidinone, 1-benzyl-3-pyrrolidone, 1-benzyl-pyrrolidin-3-one, 1-benzyl pyrrolidin-3-one, 3-pyrrolidinone, 1-phenylmethyl, benzyl-3-pyrrolidinone, pubchem10007, n-benzylpyrrolid-3-one | |
1-benzylpyrrolidin-3-one |
Specifications
775-16-6 | |
77°C to 78°C (0.01 mmHg) | |
C11H13NO | |
1-benzyl-3-pyrrolidinone, n-benzyl-3-pyrrolidinone, 1-phenylmethyl-3-pyrrolidinone, 1-benzyl-3-pyrrolidone, 1-benzyl-pyrrolidin-3-one, 1-benzyl pyrrolidin-3-one, 3-pyrrolidinone, 1-phenylmethyl, benzyl-3-pyrrolidinone, pubchem10007, n-benzylpyrrolid-3-one | |
C1CN(CC1=O)CC2=CC=CC=C2 | |
175.231 | |
175.23 | |
1-Benzyl-3-pyrrolidinone |
1.091 | |
>110°C (230°F) | |
MFCD00005342 | |
DHGMDHQNUNRMIN-UHFFFAOYSA-N | |
1-benzylpyrrolidin-3-one | |
69890 | |
98% |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 212-274-0
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only