Learn More
1-Benzyl-4-ethoxycarbonyl-3-piperidone hydrochloride, tech. 90%, Thermo Scientific™
£23.60 - £55.70
Chemical Identifiers
CAS | 52763-21-0 |
---|---|
Molecular Formula | C15H20ClNO3 |
Molecular Weight (g/mol) | 297.78 |
MDL Number | MFCD00012792 |
InChI Key | UQOMEAWPKSISII-UHFFFAOYNA-N |
Synonym | ethyl 1-benzyl-3-oxopiperidine-4-carboxylate hydrochloride, ethyl n-benzyl-3-oxo-4-piperidine-carboxylate hydrochloride, ethyl 1-benzyl-3-oxo-4-piperidinecarboxylate hydrochloride, 1-benzyl-4-ethoxycarbonyl-3-piperidone hydrochloride, ethyl 1-benzyl-3-oxo-piperidine-4-carboxylate hcl, ethyl 1-benzyl-3-oxo-piperidine-4-carboxylate hydrochloride, 1-benzyl-3-oxopiperidine-4-carboxylic acid ethyl ester hydrochloride, ethyl n-benzyl-3-oxopiperidine-4-carboxylate hydrochloride, zlchem 420, pubchem19671 |
PubChem CID | 2723880 |
IUPAC Name | ethyl 1-benzyl-3-oxopiperidine-4-carboxylate;hydrochloride |
SMILES | [H+].[Cl-].CCOC(=O)C1CCN(CC2=CC=CC=C2)CC1=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11321927
|
Thermo Scientific Alfa Aesar
L07472.03 |
1 g |
£23.60
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11331927
|
Thermo Scientific Alfa Aesar
L07472.06 |
5 g | N/A | ||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
52763-21-0 | |
297.78 | |
UQOMEAWPKSISII-UHFFFAOYNA-N | |
2723880 | |
[H+].[Cl-].CCOC(=O)C1CCN(CC2=CC=CC=C2)CC1=O |
C15H20ClNO3 | |
MFCD00012792 | |
ethyl 1-benzyl-3-oxopiperidine-4-carboxylate hydrochloride, ethyl n-benzyl-3-oxo-4-piperidine-carboxylate hydrochloride, ethyl 1-benzyl-3-oxo-4-piperidinecarboxylate hydrochloride, 1-benzyl-4-ethoxycarbonyl-3-piperidone hydrochloride, ethyl 1-benzyl-3-oxo-piperidine-4-carboxylate hcl, ethyl 1-benzyl-3-oxo-piperidine-4-carboxylate hydrochloride, 1-benzyl-3-oxopiperidine-4-carboxylic acid ethyl ester hydrochloride, ethyl n-benzyl-3-oxopiperidine-4-carboxylate hydrochloride, zlchem 420, pubchem19671 | |
ethyl 1-benzyl-3-oxopiperidine-4-carboxylate;hydrochloride |
Specifications
52763-21-0 | |
C15H20ClNO3 | |
3749159 | |
UQOMEAWPKSISII-UHFFFAOYNA-N | |
ethyl 1-benzyl-3-oxopiperidine-4-carboxylate;hydrochloride | |
2723880 | |
90% | |
1-Benzyl-4-ethoxycarbonyl-3-piperidone hydrochloride |
∽158°C (decomposition) | |
MFCD00012792 | |
ethyl 1-benzyl-3-oxopiperidine-4-carboxylate hydrochloride, ethyl n-benzyl-3-oxo-4-piperidine-carboxylate hydrochloride, ethyl 1-benzyl-3-oxo-4-piperidinecarboxylate hydrochloride, 1-benzyl-4-ethoxycarbonyl-3-piperidone hydrochloride, ethyl 1-benzyl-3-oxo-piperidine-4-carboxylate hcl, ethyl 1-benzyl-3-oxo-piperidine-4-carboxylate hydrochloride, 1-benzyl-3-oxopiperidine-4-carboxylic acid ethyl ester hydrochloride, ethyl n-benzyl-3-oxopiperidine-4-carboxylate hydrochloride, zlchem 420, pubchem19671 | |
[H+].[Cl-].CCOC(=O)C1CCN(CC2=CC=CC=C2)CC1=O | |
297.78 | |
297.78 | |
Technical |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 258-162-5
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.