Learn More
1-Boc-1,2,3,6-tetrahydropyridine, 97%, Thermo Scientific Chemicals
£22.50 - £224.00
Chemical Identifiers
CAS | 85838-94-4 |
---|---|
Molecular Formula | C10H17NO2 |
Molecular Weight (g/mol) | 183.251 |
MDL Number | MFCD04972245 |
InChI Key | SHHHRQFHCPINIB-UHFFFAOYSA-N |
Synonym | n-boc-1,2,3,6-tetrahydropyridine, tert-butyl 5,6-dihydropyridine-1 2h-carboxylate, tert-butyl 1,2,3,6-tetrahydropyridine-1-carboxylate, tert-butyl 3,6-dihydropyridine-1 2h-carboxylate, 1-boc-1,2,3,6-tetrahydro-pyridine, 1-boc-1,2,3,6-tetrahydropyridine, n-boc-1,2,5,6-tetrahydropyridine, n-tert-butoxycarbonyl-1,2,3,6-tetrahydropyridine, 1 2h-pyridinecarboxylic acid, 3,6-dihydro-, 1,1-dimethylethyl ester, 3,6-dihydro-2h-pyridine-1-carboxylic acid tert-butyl ester |
PubChem CID | 13094787 |
IUPAC Name | tert-butyl 3,6-dihydro-2H-pyridine-1-carboxylate |
SMILES | CC(C)(C)OC(=O)N1CCC=CC1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15489327
|
Thermo Scientific Alfa Aesar
H52574.MD |
250 mg |
£22.50
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15469327
|
Thermo Scientific Alfa Aesar
H52574.03 |
1 g |
£58.80
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15479327
|
Thermo Scientific Alfa Aesar
H52574.06 |
5 g |
£224.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1-Boc-1,2,3,6-tetrahydropyridine is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Boc-1,2,3,6-tetrahydropyridine is used as pharmaceutical intermediate.
Solubility
Insoluble in water. Soluble in organic solvents.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
Chemical Identifiers
85838-94-4 | |
183.251 | |
SHHHRQFHCPINIB-UHFFFAOYSA-N | |
13094787 | |
CC(C)(C)OC(=O)N1CCC=CC1 |
C10H17NO2 | |
MFCD04972245 | |
n-boc-1,2,3,6-tetrahydropyridine, tert-butyl 5,6-dihydropyridine-1 2h-carboxylate, tert-butyl 1,2,3,6-tetrahydropyridine-1-carboxylate, tert-butyl 3,6-dihydropyridine-1 2h-carboxylate, 1-boc-1,2,3,6-tetrahydro-pyridine, 1-boc-1,2,3,6-tetrahydropyridine, n-boc-1,2,5,6-tetrahydropyridine, n-tert-butoxycarbonyl-1,2,3,6-tetrahydropyridine, 1 2h-pyridinecarboxylic acid, 3,6-dihydro-, 1,1-dimethylethyl ester, 3,6-dihydro-2h-pyridine-1-carboxylic acid tert-butyl ester | |
tert-butyl 3,6-dihydro-2H-pyridine-1-carboxylate |
Specifications
85838-94-4 | |
C10H17NO2 | |
n-boc-1,2,3,6-tetrahydropyridine, tert-butyl 5,6-dihydropyridine-1 2h-carboxylate, tert-butyl 1,2,3,6-tetrahydropyridine-1-carboxylate, tert-butyl 3,6-dihydropyridine-1 2h-carboxylate, 1-boc-1,2,3,6-tetrahydro-pyridine, 1-boc-1,2,3,6-tetrahydropyridine, n-boc-1,2,5,6-tetrahydropyridine, n-tert-butoxycarbonyl-1,2,3,6-tetrahydropyridine, 1 2h-pyridinecarboxylic acid, 3,6-dihydro-, 1,1-dimethylethyl ester, 3,6-dihydro-2h-pyridine-1-carboxylic acid tert-butyl ester | |
SHHHRQFHCPINIB-UHFFFAOYSA-N | |
tert-butyl 3,6-dihydro-2H-pyridine-1-carboxylate | |
13094787 | |
97% | |
1-Boc-1,2,3,6-tetrahydropyridine |
78°C to 84°C (5mmHg) | |
MFCD04972245 | |
Insoluble in water. Soluble in organic solvents. | |
CC(C)(C)OC(=O)N1CCC=CC1 | |
183.251 | |
183.25 | |
1.029 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only