missing translation for 'onlineSavingsMsg'
Learn More
1-Boc-3-iodoazetidine, 97% Product Code.: 15419307

1-Boc-3-iodoazetidine, 97%

Product Code. 15419307
5 g
Click to view available options
5 g
This item is not returnable. View return policy

Product Code. 15419307

Brand: Thermo Scientific Alfa Aesar H52558.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 254454-54-1 | C8H14INO2 | 283.109 g/mol

1-Boc-3-iodoazetidine is used as an organic chemical synthesis intermediate.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1-Boc-3-iodoazetidine is used as an organic chemical synthesis intermediate.

Notes
Light sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Specifications

CAS 254454-54-1
Molecular Formula C8H14INO2
MDL Number MFCD09752821
Quantity 5 g
Sensitivity Light sensitive
Synonym 1-boc-3-iodoazetidine, 1-boc-3-iodo-azetidine, 1-boc-3-iodo azetidine, n-boc-3-iodoazetidine, 3-iodo-azetidine-1-carboxylic acid tert-butyl ester
InChI Key XPDIKRMPZNLBAC-UHFFFAOYSA-N
SMILES CC(C)(C)OC(=O)N1CC(C1)I
IUPAC Name tert-butyl 3-iodoazetidine-1-carboxylate
Molecular Weight (g/mol) 283.109
PubChem CID 11000522
Formula Weight 283.11
Percent Purity 97%
Chemical Name or Material 1-Boc-3-iodoazetidine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberI19X015 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH52558.06
Lot NumberA14Z025 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH52558.06
Lot NumberF21Z048 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH52558.06
Lot NumberJ13X012 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH52558.06
Lot NumberN15D032 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH52558.06
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Boc-3-iodoazetidine, 97% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.