missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Boc-3-(iodomethyl)azetidine, 95%, Thermo Scientific™
£88.60 - £131.00
Chemical Identifiers
CAS | 253176-94-2 |
---|---|
Molecular Formula | C9H16INO2 |
Molecular Weight (g/mol) | 297.14 |
MDL Number | MFCD09951816 |
InChI Key | CJVGFEARJOREHI-UHFFFAOYSA-N |
PubChem CID | 53249943 |
IUPAC Name | tert-butyl 3-(iodomethyl)azetidine-1-carboxylate |
SMILES | CC(C)(C)OC(=O)N1CC(CI)C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15423968
|
Thermo Scientific Alfa Aesar
H57500.MD |
250 mg |
£88.60
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15433968
|
Thermo Scientific Alfa Aesar
H57500.ME |
500 mg | |||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
253176-94-2 | |
297.14 | |
CJVGFEARJOREHI-UHFFFAOYSA-N | |
tert-butyl 3-(iodomethyl)azetidine-1-carboxylate |
C9H16INO2 | |
MFCD09951816 | |
53249943 | |
CC(C)(C)OC(=O)N1CC(CI)C1 |
Specifications
253176-94-2 | |
MFCD09951816 | |
CJVGFEARJOREHI-UHFFFAOYSA-N | |
tert-butyl 3-(iodomethyl)azetidine-1-carboxylate | |
53249943 | |
95% |
C9H16INO2 | |
tert-butyl 3-iodomethyl azetidine-1-carboxylate, 1-boc-3-iodomethyl azetidine, n-boc-3-iodomethyl azetidine, tert-butyl3-iodomethyl azetidine-1-carboxylate, 1-azetidinecarboxylic acid, 3-iodomethyl-, 1,1-dimethylethyl ester, acmc-209gj7, 1-n-boc-3-iodomethyl azetidine, tert-butyl-3-iodomethyl azetidin-1-carboxylate, tert-butyl 3-iodomethyl ;azetidine-1-carboxylate, tert-butyl 3-iodanylmethyl azetidine-1-carboxylate | |
CC(C)(C)OC(=O)N1CC(CI)C1 | |
297.14 | |
297.13 | |
1-Boc-3-(iodomethyl)azetidine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures