Learn More
1-Bromo-2-methylpropane, 98+%, Thermo Scientific Chemicals
£24.00 - £325.00
Chemical Identifiers
CAS | 78-77-3 |
---|---|
Molecular Formula | C4H9Br |
Molecular Weight (g/mol) | 137.02 |
InChI Key | HLVFKOKELQSXIQ-UHFFFAOYSA-N |
Synonym | isobutyl bromide, iso-butyl bromide, propane, 1-bromo-2-methyl, bromoisobutane, i-butyl bromide, 1-bromo-2-methyl-propane, unii-5oec0bw987, ccris 349, 1-bromo-2-methyl propane, bromoacetaldehydedimethylacetal |
PubChem CID | 6555 |
IUPAC Name | 1-bromo-2-methylpropane |
SMILES | CC(C)CBr |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10606404
|
Thermo Scientific Acros
168831000 |
100 mL | Glass bottle |
£24.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10062110
|
Thermo Scientific Acros
168835000 |
500 mL | Glass bottle |
£82.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11974491
|
Thermo Scientific Acros
168830025 |
2.5 L | Glass bottle |
£325.00
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
78-77-3 | |
137.02 | |
isobutyl bromide, iso-butyl bromide, propane, 1-bromo-2-methyl, bromoisobutane, i-butyl bromide, 1-bromo-2-methyl-propane, unii-5oec0bw987, ccris 349, 1-bromo-2-methyl propane, bromoacetaldehydedimethylacetal | |
1-bromo-2-methylpropane |
C4H9Br | |
HLVFKOKELQSXIQ-UHFFFAOYSA-N | |
6555 | |
CC(C)CBr |
Specifications
78-77-3 | |
Colorless | |
90.0°C to 92.0°C | |
Authentic | |
C4H9Br | |
1.4346 to 1.4366 | |
15,518 | |
isobutyl bromide, iso-butyl bromide, propane, 1-bromo-2-methyl, bromoisobutane, i-butyl bromide, 1-bromo-2-methyl-propane, unii-5oec0bw987, ccris 349, 1-bromo-2-methyl propane, bromoacetaldehydedimethylacetal | |
HLVFKOKELQSXIQ-UHFFFAOYSA-N | |
1-bromo-2-methylpropane | |
6555 | |
98+% | |
1-Bromo-2-methylpropane |
-119.0°C | |
1.2600g/mL | |
18°C | |
98% min. (GC) | |
(CH3)2CHCH2Br | |
01,126 | |
1.26 | |
Solubility in water: 0.51g/L (18°C). Other solubilities: soluble in most common organic solvents | |
CC(C)CBr | |
137.02 | |
137.02 | |
Liquid |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.
GHS P Statement
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wash face,hands and any exposed skin thoroughly after handling.
Wear protective gloves/protective clothing/eye protect
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 201-141-2
- 1-Bromo-2-methylpropane
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only