missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-4-butoxybenzene, 97%, Thermo Scientific™
£24.20 - £128.00
Chemical Identifiers
CAS | 39969-57-8 |
---|---|
Molecular Formula | C10H13BrO |
Molecular Weight (g/mol) | 229.117 |
MDL Number | MFCD00037100 |
InChI Key | BOUVKHWPQNEXTO-UHFFFAOYSA-N |
PubChem CID | 142395 |
IUPAC Name | 1-bromo-4-butoxybenzene |
SMILES | CCCCOC1=CC=C(C=C1)Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10096804
|
Thermo Scientific Maybridge
BTB09793DE |
5 g |
£24.20
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10140722
|
Thermo Scientific Maybridge
BTB09793EA |
10 g |
£40.30
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10661663
|
Thermo Scientific Maybridge
BTB09793EE |
50 g |
£128.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
39969-57-8 | |
229.117 | |
BOUVKHWPQNEXTO-UHFFFAOYSA-N | |
1-bromo-4-butoxybenzene |
C10H13BrO | |
MFCD00037100 | |
142395 | |
CCCCOC1=CC=C(C=C1)Br |
Specifications
39969-57-8 | |
100.0 | |
C10H13BrO | |
MFCD00037100 | |
BOUVKHWPQNEXTO-UHFFFAOYSA-N | |
1-bromo-4-butoxybenzene | |
142395 | |
97% |
96.5 | |
97% | |
BrC6H4O(CH2)3CH3 | |
p-bromophenyl butyl ether, benzene,1-bromo-4-butoxy, p-butoxybromobenzene, benzene, 1-bromo-4-butoxy, 4-bromo-1-butoxybenzene, 4-butoxybromobenzene, 4-n-butoxyphenyl bromide, 1-butoxy-4-bromobenzene, 4-bromophenyl butyl ether | |
CCCCOC1=CC=C(C=C1)Br | |
229.117 | |
229.12 | |
1-Bromo-4-butoxybenzene |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title