Learn More
1-Bromo-5-fluoropentane, 99%, Thermo Scientific Chemicals
£17.20 - £272.00
Chemical Identifiers
CAS | 407-97-6 |
---|---|
Molecular Formula | C5H10BrF |
Molecular Weight (g/mol) | 169.037 |
MDL Number | MFCD01709395 |
InChI Key | GMYIZICPHREVDH-UHFFFAOYSA-N |
Synonym | pentane, 1-bromo-5-fluoro, 5-fluoroamyl bromide, 5-fluoropentyl bromide, fluoropentyl bromide, 5-bromopentyl fluoride, acmc-20a2uo, 1-bromo-5-fluoro-pentane, pentane,1-bromo-5-fluoro, 4-01-00-00313 beilstein handbook reference, ksc493m6b |
PubChem CID | 120236 |
IUPAC Name | 1-bromo-5-fluoropentane |
SMILES | C(CCF)CCBr |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15412179
|
Thermo Scientific Alfa Aesar
H64806.MD |
250 mg | |||||||
15492169
|
Thermo Scientific Alfa Aesar
H64806.03 |
1 g |
£70.40
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15402179
|
Thermo Scientific Alfa Aesar
H64806.06 |
5 g |
£272.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1-Bromo-5-fluoropentane was used to prepare 18F-labeled (anilido)piperidine derivatives as ?-opioid receptor imaging agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Bromo-5-fluoropentane was used to prepare 18F-labeled (anilido)piperidine derivatives as μ-opioid receptor imaging agents.
Solubility
Soluble in methanol (in methanol). Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Chemical Identifiers
407-97-6 | |
169.037 | |
GMYIZICPHREVDH-UHFFFAOYSA-N | |
120236 | |
C(CCF)CCBr |
C5H10BrF | |
MFCD01709395 | |
pentane, 1-bromo-5-fluoro, 5-fluoroamyl bromide, 5-fluoropentyl bromide, fluoropentyl bromide, 5-bromopentyl fluoride, acmc-20a2uo, 1-bromo-5-fluoro-pentane, pentane,1-bromo-5-fluoro, 4-01-00-00313 beilstein handbook reference, ksc493m6b | |
1-bromo-5-fluoropentane |
Specifications
407-97-6 | |
1.360 g/mL | |
C5H10BrF | |
1.4406 | |
1697258 | |
Soluble in methanol (in methanol). Slightly soluble in water. | |
C(CCF)CCBr | |
169.037 | |
169.04 | |
Liquid |
Colorless | |
162°C | |
F(CH2)5Br | |
MFCD01709395 | |
pentane, 1-bromo-5-fluoro, 5-fluoroamyl bromide, 5-fluoropentyl bromide, fluoropentyl bromide, 5-bromopentyl fluoride, acmc-20a2uo, 1-bromo-5-fluoro-pentane, pentane,1-bromo-5-fluoro, 4-01-00-00313 beilstein handbook reference, ksc493m6b | |
GMYIZICPHREVDH-UHFFFAOYSA-N | |
1-bromo-5-fluoropentane | |
120236 | |
99% | |
1-Bromo-5-fluoropentane |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'rtecsNumber' : RZ9810000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only