Learn More
1-Bromonaphthalene, 97%, Thermo Scientific Chemicals
Description
1-Bromonaphthalene is used in organic synthesis and refractometry of fats. It is used in the preparation of N-aryl imidazoles and diaryl ethers. It is used to determine the refractive index of crystals and the water content of alcohols. It is utilized in the preparation of glutathione peroxidase-like antioxidant activity of diaryl diselenides. Further, it serves as a solvent for the exfoliation and dispersion of hexabenzocoronene. In addition to this, it is used as a refrigerant and a solvent molecular weight substance.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1-Bromonaphthalene is used in organic synthesis and refractometry of fats. It is used in the preparation of N-aryl imidazoles and diaryl ethers. It is used to determine the refractive index of crystals and the water content of alcohols. It is utilized in the preparation of glutathione peroxidase-like antioxidant activity of diaryl diselenides. Further, it serves as a solvent for the exfoliation and dispersion of hexabenzocoronene. In addition to this, it is used as a refrigerant and a solvent molecular weight substance.
Solubility
Miscible with chloroform, ethanol, ether, benzene and ethyl acetate. Slightly miscible with water.
Notes
Incompatible with strong oxidizing agents.

Specifications
Specifications
Melting Point | -2°C to 0°C |
Density | 1.485 |
Boiling Point | 279°C to 281°C |
Flash Point | >110°C (230°F) |
Assay Percent Range | 97% |
Refractive Index | 1.657 |
Quantity | 1000 g |
Beilstein | 1906414 |
Merck Index | 14,1425 |
Solubility Information | Miscible with chloroform,ethanol,ether,benzene and ethyl acetate. Slightly miscible with water. |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0462233 | Certificate of Analysis | 30/08/2024 | A14122.0B,A14122.18,A14122.30 |
A0460987 | Certificate of Analysis | 16/04/2024 | A14122.0B,A14122.18,A14122.30 |
A0458232 | Certificate of Analysis | 07/02/2024 | A14122.0B,A14122.18,A14122.30 |
A0456260 | Certificate of Analysis | 31/01/2024 | A14122.0B,A14122.18,A14122.30 |
A0456474 | Certificate of Analysis | 25/10/2023 | A14122.0B,A14122.18,A14122.30 |
Safety and Handling


- 1-Bromonaphthalene
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 1
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- H410-Very toxic to aquatic life with long lasting effects.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : 1-Bromonaphthalene
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.