missing translation for 'onlineSavingsMsg'
Learn More
1-Bromopentadecane, 97%, Thermo Scientific Chemicals Product Code.: 11351987

1-Bromopentadecane, 97%, Thermo Scientific Chemicals

Product Code. 11351987
10 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 11351987

Brand: Thermo Scientific Alfa Aesar L07618.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1-Bromopentadecane has been used as regent in the synthesis of (S)-eicos-(4E)-en-1-yn-3-ol, a bioactive component of the marine sponge Cribrochalina vasculum.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromopentadecane has been used as regent in the synthesis of (S)-eicos-(4E)-en-1-yn-3-ol, a bioactive component of the marine sponge Cribrochalina vasculum.

Solubility
Not miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents. Incompatible materials are oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629-72-1
Molecular Formula C15H31Br
Molecular Weight (g/mol) 291.32
MDL Number MFCD00000229
InChI Key JKOTZBXSNOGCIF-UHFFFAOYSA-N
Synonym pentadecyl bromide, pentadecane, 1-bromo, 1-bromo pentadecane, n-pentadecyl bromide, pentadecane,1-bromo, 1-bromopentadecane, acmc-209nb8, dsstox_cid_29296, dsstox_rid_83414, dsstox_gsid_49339Show MoreShow Less
PubChem CID 12394
IUPAC Name 1-bromopentadecane
SMILES CCCCCCCCCCCCCCCBr

Specifications

Melting Point 18°C to 19°C
Density 1.005
Boiling Point 159°C to 160°C (5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.4608
Quantity 10 g
Beilstein 1744553
Solubility Information Not miscible in water.
Formula Weight 291.32
Percent Purity 97%
Chemical Name or Material 1-Bromopentadecane
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10250103 Certificate TypeCertificate of Analysis Date25/03/2024 Catalog NumberL07618.18,L07618.09
Lot NumberJ13X016 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL07618.18,L07618.09
Lot Number10200626 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL07618.18,L07618.09
Lot Number10148546 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL07618.18,L07618.09
Lot Number10208392 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL07618.18,L07618.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Bromopentadecane, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.