Learn More
1-Bromopentadecane, 97%, Thermo Scientific Chemicals
Description
1-Bromopentadecane has been used as regent in the synthesis of (S)-eicos-(4E)-en-1-yn-3-ol, a bioactive component of the marine sponge Cribrochalina vasculum.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1-Bromopentadecane has been used as regent in the synthesis of (S)-eicos-(4E)-en-1-yn-3-ol, a bioactive component of the marine sponge Cribrochalina vasculum.
Solubility
Not miscible in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents. Incompatible materials are oxidizing agents.

Specifications
Specifications
Melting Point | 18°C to 19°C |
Density | 1.005 |
Boiling Point | 159°C to 160°C (5 mmHg) |
Flash Point | >110°C (230°F) |
Refractive Index | 1.4608 |
Quantity | 50 g |
Beilstein | 1744553 |
Solubility Information | Not miscible in water. |
Formula Weight | 291.32 |
Percent Purity | 97% |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10250103 | Certificate of Analysis | 25/03/2024 | L07618.18,L07618.09 |
10246531 | Certificate of Analysis | 04/05/2023 | L07618.18 |
J13X016 | Certificate of Analysis | 08/12/2021 | L07618.18,L07618.09 |
10200626 | Certificate of Analysis | 08/12/2021 | L07618.18,L07618.09 |
10148546 | Certificate of Analysis | 08/12/2021 | L07618.18,L07618.09 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.