missing translation for 'onlineSavingsMsg'
Learn More
1-Butanol, Thermo Scientific Chemicals Product Code.: 10275064

1-Butanol, Thermo Scientific Chemicals

Product Code. 10275064
1 L, Glass Bottle
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
This item is not returnable. View return policy

Product Code. 10275064

Brand: Thermo Scientific Acros 393750010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For HPLC
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-36-3
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
Synonym 1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropaneShow MoreShow Less
PubChem CID 263
ChEBI CHEBI:28885
IUPAC Name butan-1-ol
SMILES CCCCO

Specifications

Melting Point -89.0°C
Absorbance 0.046 max. (1cm cell vs HLPC−grade water) at 245nm, 0.301 max. (1cm cell vs HPLC−grade water) at 220nm, 0.699 max. (1 cm cell vs HPLC−grade water) at 210nm
Density 0.8100g/mL
Boiling Point 117.6°C
Quantity 1 L
Flash Point 35°C
Packaging Glass Bottle
Refractive Index 1.3980 to 1.4000
Specific Gravity 0.81
Linear Formula CH3(CH2)3OH
Viscosity 2.95 mPa.s (20°C)
Solubility Information Solubility in water: 80g/L (20°C). Other solubilities: miscible with ethanol, ether and many other, organic solvents
Formula Weight 74.12
Grade HPLC
Chemical Name or Material 1-Butanol, For HPLC
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465288 Certificate TypeCertificate of Analysis Date07/08/2024 Catalog Number393750025,393750010
Lot NumberA0459073 Certificate TypeCertificate of Analysis Date08/01/2024 Catalog Number393750025,393750010
Lot NumberA0455215 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number393750025,393750010
Lot NumberA0450209 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number393750025,393750010
Lot NumberA0450210 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number393750025,393750010
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1-Butanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 1-Butanol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Butanol, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.