missing translation for 'onlineSavingsMsg'
Learn More
1-Butenyl ethyl ether, cis + trans, 98%, Thermo Scientific Chemicals Product Code.: 11332689

1-Butenyl ethyl ether, cis + trans, 98%, Thermo Scientific Chemicals

Product Code. 11332689
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11332689

Brand: Thermo Scientific Alfa Aesar H26720.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 929-05-5
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.161
MDL Number MFCD03427069
InChI Key AQTYNINXYJFSHD-AATRIKPKSA-N
Synonym e-1-ethoxybut-1-ene, 1-butenyl ethyl ether, 1e-1-ethoxybut-1-ene, 1-ethoxybut-1-ene, 1-ethoxy-1-butene, cis-1-butenyl ethyl ether, trans-1-butenyl ethyl ether, 1-butene, 1-ethoxy-, e, trans-1-ethoxy-1-butene, 1e-1-butenyl ethyl ether #Show MoreShow Less
PubChem CID 5364930
IUPAC Name (E)-1-ethoxybut-1-ene
SMILES CCC=COCC

Specifications

Boiling Point 94°C to 95°C
Flash Point 6°C (43°F)
Linear Formula CH3CH2CH=CHOCH2CH3
Quantity 5 g
UN Number UN3271
Formula Weight 100.16
Percent Purity 98%
Chemical Name or Material 1-Butenyl ethyl ether, Cis + Trans

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10210648 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH26720.14,H26720.06,H26720.22
Lot Number10131506 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH26720.14,H26720.06,H26720.22
Lot Number10207740 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH26720.14,H26720.06,H26720.22
Lot Number10181566 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH26720.14,H26720.06,H26720.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Butenyl ethyl ether, cis + trans, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.