Learn More
1-Chloro-2-methylpropyl chloroformate, 98%, Thermo Scientific Chemicals
£35.10 - £121.00
Chemical Identifiers
CAS | 92600-11-8 |
---|---|
Molecular Formula | C5H8Cl2O2 |
Molecular Weight (g/mol) | 171.02 |
MDL Number | MFCD00012387 |
InChI Key | BOGLZTPABVHWDD-BYPYZUCNSA-N |
Synonym | 1-chloro-2-methylpropyl chloroformate, acmc-20aow3, 1-chloro-2-methylpropylchloroformate, 1-chloro-2-methylpropyl carbonochloridate, chloro 1-chloro-2-methylpropoxy methanone, carbonochloridic acid,1-chloro-2-methylpropyl ester |
PubChem CID | 2733326 |
SMILES | CC(C)[C@@H](Cl)OC(Cl)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10277660
|
Thermo Scientific Acros
314470010 |
1 g | Glass bottle |
£35.10
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10781345
|
Thermo Scientific Acros
314470050 |
5 g | Glass bottle |
£121.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagentChemical Identifiers
92600-11-8 | |
171.02 | |
BOGLZTPABVHWDD-BYPYZUCNSA-N | |
2733326 |
C5H8Cl2O2 | |
MFCD00012387 | |
1-chloro-2-methylpropyl chloroformate, acmc-20aow3, 1-chloro-2-methylpropylchloroformate, 1-chloro-2-methylpropyl carbonochloridate, chloro 1-chloro-2-methylpropoxy methanone, carbonochloridic acid,1-chloro-2-methylpropyl ester | |
CC(C)[C@@H](Cl)OC(Cl)=O |
Specifications
92600-11-8 | |
100.0 | |
1.1700g/mL | |
Authentic | |
C5H8Cl2O2 | |
1.4310 to 1.433 | |
1.17 | |
BOGLZTPABVHWDD-BYPYZUCNSA-N | |
(1R)-1-chloro-2-methylpropyl carbonochloridate | |
2733326 | |
98% | |
1-chloro-2-methylpropyl chloroformate |
97.5 | |
Colorless | |
53°C | |
98% | |
ClCO2CH(Cl)CH(CH3)2 | |
MFCD00012387 | |
1-chloro-2-methylpropyl chloroformate, acmc-20aow3, 1-chloro-2-methylpropylchloroformate, 1-chloro-2-methylpropyl carbonochloridate, chloro 1-chloro-2-methylpropoxy methanone, carbonochloridic acid,1-chloro-2-methylpropyl ester | |
CC(C)[C@@H](Cl)OC(Cl)=O | |
171.02 | |
171.02 | |
Liquid |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Harmful if inhaled.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Flammable liquid and vapor.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and us
GHS Signal Word: Danger
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H226-Flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H331-Toxic if inhaled.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only