Learn More
1-Ethynyl-1-cyclohexanol, 99+%, Thermo Scientific Chemicals
£24.10 - £338.00
Chemical Identifiers
CAS | 78-27-3 |
---|---|
Molecular Formula | C8H12O |
Molecular Weight (g/mol) | 124.18 |
MDL Number | MFCD00003858 |
InChI Key | QYLFHLNFIHBCPR-UHFFFAOYSA-N |
Synonym | 1-ethynyl-1-cyclohexanol, 1-ethynylcyclohexanol, ethynylcyclohexanol, cyclohexanol, 1-ethynyl, 1-hydroxycyclohexyl ethyne, 1-ethynyl-1-hydroxycyclohexane, 1-ethynyl cyclohexanol, 1-ethynyl-cyclohexanol, unii-6rv04025eh, 1-ethynyl-cyclohexan-1-ol |
PubChem CID | 6525 |
IUPAC Name | 1-ethynylcyclohexan-1-ol |
SMILES | C#CC1(CCCCC1)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10569600
|
Thermo Scientific Acros
119101000 |
100 g | Glass bottle |
£24.10
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10326510
|
Thermo Scientific
119105000 |
500 g | Glass bottle |
£80.30
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11974641
|
Thermo Scientific Acros
119100025 |
2.5 kg | Glass bottle |
£338.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
78-27-3 | |
124.18 | |
QYLFHLNFIHBCPR-UHFFFAOYSA-N | |
6525 | |
C#CC1(CCCCC1)O |
C8H12O | |
MFCD00003858 | |
1-ethynyl-1-cyclohexanol, 1-ethynylcyclohexanol, ethynylcyclohexanol, cyclohexanol, 1-ethynyl, 1-hydroxycyclohexyl ethyne, 1-ethynyl-1-hydroxycyclohexane, 1-ethynyl cyclohexanol, 1-ethynyl-cyclohexanol, unii-6rv04025eh, 1-ethynyl-cyclohexan-1-ol | |
1-ethynylcyclohexan-1-ol |
Specifications
78-27-3 | |
100.0 | |
Colorless to Yellow | |
180.0°C | |
Authentic | |
C8H12O | |
1.4810 to 1.4840 | |
06,II,1 | |
0.9763 | |
Solubility in water: 24.5g/L (20°C). Other solubilities: soluble in most organic solvents, soluble in diethyl ether, alcohols, esters, ketone, and chlorinated hydrocarbons | |
C#CC1(CCCCC1)O | |
124.18 | |
124.18 | |
Liquid After Melting |
99.0 | |
30.0°C to 32.0°C | |
0.9763g/mL | |
73°C | |
99% min. (GC) | |
HC≡CC6H10OH | |
MFCD00003858 | |
10,381 | |
1-ethynyl-1-cyclohexanol, 1-ethynylcyclohexanol, ethynylcyclohexanol, cyclohexanol, 1-ethynyl, 1-hydroxycyclohexyl ethyne, 1-ethynyl-1-hydroxycyclohexane, 1-ethynyl cyclohexanol, 1-ethynyl-cyclohexanol, unii-6rv04025eh, 1-ethynyl-cyclohexan-1-ol | |
QYLFHLNFIHBCPR-UHFFFAOYSA-N | |
1-ethynylcyclohexan-1-ol | |
6525 | |
99+% | |
1-Ethynyl-1-cyclohexanol, 99+% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
Toxic in contact with skin.
Causes skin irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
Take off contaminated clothing a
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 201-100-9
missing translation for 'rtecsNumber' : GV9100000
missing translation for 'tsca' : TSCA
- 1-Ethynyl-1-cyclohexanol
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H311-Toxic in contact with skin.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P362-Take off contaminated clothing.
RUO – Research Use Only