Learn More
1-Ethynylcyclopentanol, 98+%, Thermo Scientific Chemicals
£37.99 - £445.00
Chemical Identifiers
CAS | 17356-19-3 |
---|---|
Molecular Formula | C7H10O |
Molecular Weight (g/mol) | 110.156 |
MDL Number | MFCD00001365 |
InChI Key | LQMDOONLLAJAPZ-UHFFFAOYSA-N |
Synonym | 1-ethynylcyclopentanol, cyclopentanol, 1-ethynyl, 1-ethynyl-1-cyclopentanol, cyclopentyl ethynyl carbinol, 1-ethynyl-cyclopentan-1-ol, acmc-20abvz, 1-ethinylcyclopentanol, 1-ethynyl-cyclopentanol, cyclopentanol, ethynyl, 1-ethynylcyclopentanol ? |
PubChem CID | 87074 |
IUPAC Name | 1-ethynylcyclopentan-1-ol |
SMILES | C#CC1(CCCC1)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11476316
|
Thermo Scientific Alfa Aesar
B23109.06 |
5 g |
£37.99
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11486316
|
Thermo Scientific Alfa Aesar
B23109.14 |
25 g |
£145.73
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11496316
|
Thermo Scientific Alfa Aesar
B23109.22 |
100 g | N/A | ||||||
Description
1-Ethynylcyclopentanol is used in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Ethynylcyclopentanol is used in organic synthesis.
Solubility
Insoluble in water.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Chemical Identifiers
17356-19-3 | |
110.156 | |
LQMDOONLLAJAPZ-UHFFFAOYSA-N | |
87074 | |
C#CC1(CCCC1)O |
C7H10O | |
MFCD00001365 | |
1-ethynylcyclopentanol, cyclopentanol, 1-ethynyl, 1-ethynyl-1-cyclopentanol, cyclopentyl ethynyl carbinol, 1-ethynyl-cyclopentan-1-ol, acmc-20abvz, 1-ethinylcyclopentanol, 1-ethynyl-cyclopentanol, cyclopentanol, ethynyl, 1-ethynylcyclopentanol ? | |
1-ethynylcyclopentan-1-ol |
Specifications
17356-19-3 | |
0.962 | |
48°C (118°F) | |
C7H10O | |
MFCD00001365 | |
1924167 | |
Insoluble in water. | |
C#CC1(CCCC1)O | |
110.156 | |
110.16 | |
1-Ethynylcyclopentanol |
23°C to 27°C | |
156°C to 159°C | |
Irritating | |
1.474 | |
UN1325 | |
1-ethynylcyclopentanol, cyclopentanol, 1-ethynyl, 1-ethynyl-1-cyclopentanol, cyclopentyl ethynyl carbinol, 1-ethynyl-cyclopentan-1-ol, acmc-20abvz, 1-ethinylcyclopentanol, 1-ethynyl-cyclopentanol, cyclopentanol, ethynyl, 1-ethynylcyclopentanol ? | |
LQMDOONLLAJAPZ-UHFFFAOYSA-N | |
1-ethynylcyclopentan-1-ol | |
87074 | |
≥98% |
Safety and Handling
GHS H Statement
H228-H302-H319-H335
Flammable solid.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P240-P241-P261-P264b-P270-P271-P280-P301+P312-P304+P340-P305+P351+P338-P312-P330-P370+P378q-P501c
H228-H302-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 241-385-7
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only