missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Fmoc-piperidine-4-carboxylic acid, 98%, Thermo Scientific Chemicals
£13.00 - £286.00
Chemical Identifiers
CAS | 148928-15-8 |
---|---|
Molecular Formula | C21H21NO4 |
Molecular Weight (g/mol) | 351.402 |
MDL Number | MFCD00273475 |
InChI Key | OJYOOHSQOIDDOO-UHFFFAOYSA-N |
Synonym | fmoc-inp-oh, fmoc-isonipecotic acid, 1-fmoc-piperidine-4-carboxylic acid, 1-9h-fluoren-9-ylmethoxy carbonyl piperidine-4-carboxylic acid, n-fmoc-isonipecotic acid, fmoc-piperidine-4-carboxylic acid, n-fmoc-4-piperidinecarboxylic acid, 1,4-piperidinedicarboxylicacid, 1-9h-fluoren-9-ylmethyl ester, n-9-fluorenylmethyloxycarbonyl-isonipecotic acid, 1-9h-fluoren-9-ylmethoxycarbonyl piperidine-4-carboxylic acid |
PubChem CID | 2736490 |
IUPAC Name | 1-(9H-fluoren-9-ylmethoxycarbonyl)piperidine-4-carboxylic acid |
SMILES | C1CN(CCC1C(=O)O)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15430879
|
Thermo Scientific Alfa Aesar
H63915.03 |
1 g | |||||||
15440879
|
Thermo Scientific Alfa Aesar
H63915.06 |
5 g |
£79.60
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15450879
|
Thermo Scientific Alfa Aesar
H63915.14 |
25 g |
£286.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
148928-15-8 | |
351.402 | |
OJYOOHSQOIDDOO-UHFFFAOYSA-N | |
2736490 | |
C1CN(CCC1C(=O)O)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24 |
C21H21NO4 | |
MFCD00273475 | |
fmoc-inp-oh, fmoc-isonipecotic acid, 1-fmoc-piperidine-4-carboxylic acid, 1-9h-fluoren-9-ylmethoxy carbonyl piperidine-4-carboxylic acid, n-fmoc-isonipecotic acid, fmoc-piperidine-4-carboxylic acid, n-fmoc-4-piperidinecarboxylic acid, 1,4-piperidinedicarboxylicacid, 1-9h-fluoren-9-ylmethyl ester, n-9-fluorenylmethyloxycarbonyl-isonipecotic acid, 1-9h-fluoren-9-ylmethoxycarbonyl piperidine-4-carboxylic acid | |
1-(9H-fluoren-9-ylmethoxycarbonyl)piperidine-4-carboxylic acid |
Specifications
148928-15-8 | |
White | |
MFCD00273475 | |
fmoc-inp-oh, fmoc-isonipecotic acid, 1-fmoc-piperidine-4-carboxylic acid, 1-9h-fluoren-9-ylmethoxy carbonyl piperidine-4-carboxylic acid, n-fmoc-isonipecotic acid, fmoc-piperidine-4-carboxylic acid, n-fmoc-4-piperidinecarboxylic acid, 1,4-piperidinedicarboxylicacid, 1-9h-fluoren-9-ylmethyl ester, n-9-fluorenylmethyloxycarbonyl-isonipecotic acid, 1-9h-fluoren-9-ylmethoxycarbonyl piperidine-4-carboxylic acid | |
C1CN(CCC1C(=O)O)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24 | |
351.402 | |
351.4 | |
Powder |
∼190°C (decomposition) | |
C21H21NO4 | |
7722550 | |
OJYOOHSQOIDDOO-UHFFFAOYSA-N | |
1-(9H-fluoren-9-ylmethoxycarbonyl)piperidine-4-carboxylic acid | |
2736490 | |
98% | |
1-Fmoc-piperidine-4-carboxylic acid |
Safety and Handling
missing translation for 'tsca' : No
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only