Learn More
1-Hexadecylamine, tech. 90%, remainder mainly 1-octadecylamine, Thermo Scientific Chemicals
£18.00 - £62.70
Chemical Identifiers
CAS | 143-27-1 |
---|---|
Molecular Formula | C16H35N |
Molecular Weight (g/mol) | 241.46 |
MDL Number | MFCD00008158 |
InChI Key | FJLUATLTXUNBOT-UHFFFAOYSA-N |
Synonym | hexadecylamine, 1-hexadecylamine, cetylamine, 1-hexadecanamine, 1-aminohexadecane, n-hexadecylamine, hexyldecylamine, palmitamine, palmitylamine, n-cetylamine |
PubChem CID | 8926 |
IUPAC Name | hexadecan-1-amine |
SMILES | CCCCCCCCCCCCCCCCN |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11448740
|
Thermo Scientific Alfa Aesar
B22459.14 |
25 g |
£18.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11460627
|
Thermo Scientific Alfa Aesar
B22459.30 |
250 g |
£23.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11450627
|
Thermo Scientific Alfa Aesar
B22459.0B |
1000 g |
£62.70
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1-Hexadecylamine is used for producing resin, senior detergent, anti-caking agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Hexadecylamine is used for producing resin, senior detergent, anti-caking agent.
Solubility
Insoluble in water.
Notes
Incompatible with oxidizing agents, acids.
Chemical Identifiers
143-27-1 | |
241.46 | |
FJLUATLTXUNBOT-UHFFFAOYSA-N | |
8926 | |
CCCCCCCCCCCCCCCCN |
C16H35N | |
MFCD00008158 | |
hexadecylamine, 1-hexadecylamine, cetylamine, 1-hexadecanamine, 1-aminohexadecane, n-hexadecylamine, hexyldecylamine, palmitamine, palmitylamine, n-cetylamine | |
hexadecan-1-amine |
Specifications
143-27-1 | |
0.813 | |
140°C (284°F) | |
C16H35N | |
MFCD00008158 | |
1634065 | |
Insoluble in water. | |
CCCCCCCCCCCCCCCCN | |
241.46 | |
241.46 | |
Technical | |
1-Hexadecylamine |
42°C to 47°C | |
144°C (2 mmHg) | |
90% | |
1.4496 | |
UN3259 | |
hexadecylamine, 1-hexadecylamine, cetylamine, 1-hexadecanamine, 1-aminohexadecane, n-hexadecylamine, hexyldecylamine, palmitamine, palmitylamine, n-cetylamine | |
FJLUATLTXUNBOT-UHFFFAOYSA-N | |
hexadecan-1-amine | |
8926 | |
90% | |
remainder mainly 1-octadecylamine |
Safety and Handling
GHS H Statement
H314-H302
Causes severe skin burns and eye damage.
Harmful if swallowed.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II
missing translation for 'einecsNumber' : 205-596-8
missing translation for 'rtecsNumber' : ML9100000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only