missing translation for 'onlineSavingsMsg'
Learn More
1-Iodo-4-nitrobenzene, 98+%, Thermo Scientific Chemicals Product Code.: 11406826

1-Iodo-4-nitrobenzene, 98+%, Thermo Scientific Chemicals

Product Code. 11406826
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11406826

Brand: Thermo Scientific Alfa Aesar B23950.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1-Iodo-4-nitrobenzene is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Iodo-4-nitrobenzene is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. It is sensitive to light. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 636-98-6
Molecular Formula C6H4INO2
Molecular Weight (g/mol) 249.007
MDL Number MFCD00007299
InChI Key SCCCFNJTCDSLCY-UHFFFAOYSA-N
Synonym benzene, 1-iodo-4-nitro, p-iodonitrobenzene, 4-iodonitrobenzene, p-nitroiodobenzene, 4-nitroiodobenzene, 4-iodo-nitrobenzene, p-nitrophenyl iodide, 1-iodo-4-nitro-benzene, 4-iodo-1-nitrobenzene, 4-nitro-1-iodobenzeneShow MoreShow Less
PubChem CID 12495
IUPAC Name 1-iodo-4-nitrobenzene
SMILES C1=CC(=CC=C1[N+](=O)[O-])I

Specifications

Melting Point 172°C to 177°C
Boiling Point 288°C to 290°C
Quantity 5 g
Beilstein 1100378
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 249.01
Percent Purity ≥98%
Chemical Name or Material 1-Iodo-4-nitrobenzene

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10251950 Certificate TypeCertificate of Analysis Date07/08/2024 Catalog NumberB23950.06,B23950.14,B23950.22
Lot Number10239248 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberB23950.06,B23950.14,B23950.22
Lot Number10194219 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB23950.06,B23950.14,B23950.22
Lot Number10199173 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB23950.06,B23950.14,B23950.22
Lot Number10223252 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB23950.06,B23950.14,B23950.22
compliance-icons
Product Identifier
  • 1-Iodo-4-nitrobenzene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 1-Iodo-4-nitrobenzene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Iodo-4-nitrobenzene, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.