missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methyl-1H-imidazole-4-carbaldehyde, 95%, Thermo Scientific™
£164.00 - £385.00
Chemical Identifiers
CAS | 17289-26-8 |
---|---|
Molecular Formula | C5H6N2O |
Molecular Weight (g/mol) | 110.116 |
MDL Number | MFCD03411957 |
InChI Key | CQZXDIHVSPZIGF-UHFFFAOYSA-N |
PubChem CID | 573419 |
IUPAC Name | 1-methylimidazole-4-carbaldehyde |
SMILES | CN1C=C(N=C1)C=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10381463
|
Thermo Scientific Maybridge
CC03604CB |
250 mg | Amber Glass Bottle |
£164.00
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10671075
|
Thermo Scientific Maybridge
CC03604DA |
1 g | Amber Glass Bottle |
£385.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
17289-26-8 | |
110.116 | |
CQZXDIHVSPZIGF-UHFFFAOYSA-N | |
1-methylimidazole-4-carbaldehyde |
C5H6N2O | |
MFCD03411957 | |
573419 | |
CN1C=C(N=C1)C=O |
Specifications
17289-26-8 | |
100.0 | |
C5H6N2O | |
1-methyl-1h-imidazole-4-carbaldehyde, 1-methylimidazole-4-carboxaldehyde, 1-methyl-1h-imidazole-4-carboxaldehyde, 1h-imidazole-4-carboxaldehyde, 1-methyl, methyl-1h-imidazole-4-carbaldehyde, 1-methylimidazol-4-carboxaldehyde, 4-formyl-1-methyl-1h-imidazole, 1-methyl-4-formyl-imidazole, 1-methyl-1h-imidazole-4-carbaldehyde #, 1h-imidazole-4-carboxaldehyde,1-methyl | |
CN1C=C(N=C1)C=O | |
110.116 | |
110.12 | |
1-Methyl-1H-imidazole-4-carbaldehyde |
94.5 | |
95% | |
MFCD03411957 | |
CQZXDIHVSPZIGF-UHFFFAOYSA-N | |
1-methylimidazole-4-carbaldehyde | |
573419 | |
95% |
Safety and Handling
GHS P Statement Causes skin irritation. Harmful if swallowed. May cause respiratory irritation. Causes serious eye irritation.
GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protec
Warning
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title