missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1-methyl-5-phenyl-1H-pyrazol-4-yl)methanol, Thermo Scientific™
£34.55 - £220.00
Chemical Identifiers
CAS | 499785-47-6 |
---|---|
Molecular Formula | C11H12N2O |
Molecular Weight (g/mol) | 188.23 |
MDL Number | MFCD04115390 |
InChI Key | ZVGZXURABHDVPS-UHFFFAOYSA-N |
Synonym | 1-methyl-5-phenyl-1h-pyrazol-4-yl methanol, 1-methyl-5-phenylpyrazol-4-yl methanol, 1-methyl-5-phenylpyrazol-4-yl methan-1-ol, bck |
PubChem CID | 2795476 |
SMILES | CN1N=CC(CO)=C1C1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10772483
|
Thermo Scientific Maybridge
CC28109CB |
250 mg |
£34.55
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10079431
|
Thermo Scientific Maybridge
CC28109DA |
1 g |
£114.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10097004
|
Thermo Scientific Maybridge
CC28109DE |
5 g |
£220.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
499785-47-6 | |
188.23 | |
ZVGZXURABHDVPS-UHFFFAOYSA-N | |
2795476 |
C11H12N2O | |
MFCD04115390 | |
1-methyl-5-phenyl-1h-pyrazol-4-yl methanol, 1-methyl-5-phenylpyrazol-4-yl methanol, 1-methyl-5-phenylpyrazol-4-yl methan-1-ol, bck | |
CN1N=CC(CO)=C1C1=CC=CC=C1 |
Specifications
499785-47-6 | |
MFCD04115390 | |
ZVGZXURABHDVPS-UHFFFAOYSA-N | |
(1-methyl-5-phenyl-1H-pyrazol-4-yl)methanol | |
2795476 | |
97% |
C11H12N2O | |
1-methyl-5-phenyl-1h-pyrazol-4-yl methanol, 1-methyl-5-phenylpyrazol-4-yl methanol, 1-methyl-5-phenylpyrazol-4-yl methan-1-ol, bck | |
CN1N=CC(CO)=C1C1=CC=CC=C1 | |
188.23 | |
188.23 | |
(1-methyl-5-phenyl-1H-pyrazol-4-yl)methanol |