missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methylpiperazine, 99%, Thermo Scientific Chemicals
£18.90 - £380.00
Chemical Identifiers
CAS | 109-01-3 |
---|---|
Molecular Formula | C5H12N2 |
Molecular Weight (g/mol) | 100.16 |
InChI Key | PVOAHINGSUIXLS-UHFFFAOYSA-N |
Synonym | n-methylpiperazine, piperazine, 1-methyl, 4-methylpiperazine, 1-methyl-piperazine, n-methyl piperazine, 1-methyl piperazine, methylpiperazine, n-methyl-piperazine, n-methylpiperazin, 1-methylpyperazine |
PubChem CID | 53167 |
IUPAC Name | 1-methylpiperazine |
SMILES | CN1CCNCC1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10417940
|
Thermo Scientific Acros
156910050 |
5 g | Glass bottle |
£18.90
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10427940
|
Thermo Scientific Acros
156911000 |
100 g | Glass bottle |
£23.70
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10103020
|
Thermo Scientific Acros
156915000 |
500 g | Glass bottle |
£83.10
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11926521
|
Thermo Scientific Acros
156910025 |
2.5 kg | Metal Drum |
£380.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
109-01-3 | |
100.16 | |
n-methylpiperazine, piperazine, 1-methyl, 4-methylpiperazine, 1-methyl-piperazine, n-methyl piperazine, 1-methyl piperazine, methylpiperazine, n-methyl-piperazine, n-methylpiperazin, 1-methylpyperazine | |
1-methylpiperazine |
C5H12N2 | |
PVOAHINGSUIXLS-UHFFFAOYSA-N | |
53167 | |
CN1CCNCC1 |
Specifications
109-01-3 | |
0.9030g/mL | |
42°C | |
98.5% min. (GC) | |
1.4645 to 1.4665 | |
n-methylpiperazine, piperazine, 1-methyl, 4-methylpiperazine, 1-methyl-piperazine, n-methyl piperazine, 1-methyl piperazine, methylpiperazine, n-methyl-piperazine, n-methylpiperazin, 1-methylpyperazine | |
PVOAHINGSUIXLS-UHFFFAOYSA-N | |
1-methylpiperazine | |
53167 | |
100.16 | |
Liquid |
-6.0°C | |
138.0°C | |
Authentic | |
C5H12N2 | |
0.903 | |
Solubility in water: soluble. Other solubilities: soluble in ethanol and aceton | |
CN1CCNCC1 | |
100.16 | |
4 mPa.s (20°C) | |
99% | |
1-Methylpiperazine |
Safety and Handling
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-639-5
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 1-Methylpiperazine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only