Learn More
1-n-Butyl-3-methylimidazolium dicyanamide, 97%, Thermo Scientific Chemicals
£88.00 - £679.00
Chemical Identifiers
CAS | 448245-52-1 |
---|---|
Molecular Formula | C10H15N5 |
Molecular Weight (g/mol) | 205.265 |
MDL Number | MFCD07784457 |
InChI Key | ICIVTHOGIQHZRY-UHFFFAOYSA-N |
Synonym | 1-butyl-3-methylimidazolium dicyanamide, 1-butyl-3-methylimidazolium dicyamide, 1-butyl-3-methylimidazolium; dicyanoazanide, bmim dca, dsstox_cid_27886, dsstox_rid_82639, dsstox_gsid_47910, c4mim; dicyanoazanide, 1-butyl-3-methyl-1h-imidazol-3-ium cyanoimino methylidene azanide, 1-butyl-3-methylimidazolium; cyanoimino methylidene azanide |
PubChem CID | 11171745 |
IUPAC Name | 1-butyl-3-methylimidazol-3-ium;cyanoiminomethylideneazanide |
SMILES | CCCCN1C=C[N+](=C1)C.C(=[N-])=NC#N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15485408
|
Thermo Scientific Alfa Aesar
H59175.06 |
5 g |
£88.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15495408
|
Thermo Scientific Alfa Aesar
H59175.18 |
50 g |
£679.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
448245-52-1 | |
205.265 | |
ICIVTHOGIQHZRY-UHFFFAOYSA-N | |
11171745 | |
CCCCN1C=C[N+](=C1)C.C(=[N-])=NC#N |
C10H15N5 | |
MFCD07784457 | |
1-butyl-3-methylimidazolium dicyanamide, 1-butyl-3-methylimidazolium dicyamide, 1-butyl-3-methylimidazolium; dicyanoazanide, bmim dca, dsstox_cid_27886, dsstox_rid_82639, dsstox_gsid_47910, c4mim; dicyanoazanide, 1-butyl-3-methyl-1h-imidazol-3-ium cyanoimino methylidene azanide, 1-butyl-3-methylimidazolium; cyanoimino methylidene azanide | |
1-butyl-3-methylimidazol-3-ium;cyanoiminomethylideneazanide |
Specifications
448245-52-1 | |
C10H15N5 | |
Moisture sensitive | |
ICIVTHOGIQHZRY-UHFFFAOYSA-N | |
1-butyl-3-methylimidazol-3-ium;cyanoiminomethylideneazanide | |
11171745 | |
97% | |
1-n-Butyl-3-methylimidazolium dicyanamide |
Yellow | |
MFCD07784457 | |
1-butyl-3-methylimidazolium dicyanamide, 1-butyl-3-methylimidazolium dicyamide, 1-butyl-3-methylimidazolium; dicyanoazanide, bmim dca, dsstox_cid_27886, dsstox_rid_82639, dsstox_gsid_47910, c4mim; dicyanoazanide, 1-butyl-3-methyl-1h-imidazol-3-ium cyanoimino methylidene azanide, 1-butyl-3-methylimidazolium; cyanoimino methylidene azanide | |
CCCCN1C=C[N+](=C1)C.C(=[N-])=NC#N | |
205.265 | |
205.26 | |
Liquid |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only