missing translation for 'onlineSavingsMsg'
Learn More
1-Octanethiol, 97%, Thermo Scientific Chemicals Product Code.: 10685972

1-Octanethiol, 97%, Thermo Scientific Chemicals

Product Code. 10685972
250 g, Glass bottle
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
Plastic Bottle
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 10685972

Brand: Thermo Scientific Acros 157482500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-88-6
Molecular Formula C8H18S
Molecular Weight (g/mol) 146.29
MDL Number MFCD00004912
InChI Key KZCOBXFFBQJQHH-UHFFFAOYSA-N
Synonym 1-octanethiol, 1-mercaptooctane, n-octyl mercaptan, octyl mercaptan, octanethiol, octylthiol, 1-octylthiol, n-octanethiol, octylmercaptan, n-octylthiolShow MoreShow Less
PubChem CID 8144
IUPAC Name octane-1-thiol
SMILES CCCCCCCCS

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -49.0°C
Color Colorless
Density 0.8400g/mL
Boiling Point 197.0°C to 200.0°C
Flash Point 68°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)7SH
Refractive Index 1.4525 to 1.4545
Quantity 250 g
Specific Gravity 0.84
Formula Weight 146.29
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Octanethiol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0444037 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number157480020,157480010,157482500,157480025
Lot NumberA0440000 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number157480020,157480010,157482500,157480025
Lot NumberA0456793 Certificate TypeCertificate of Analysis Date10/10/2023 Catalog Number157480020,157480010,157482500,157480025
Lot NumberA0451111 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number157480020,157480010,157482500,157480025
Lot NumberA0352858 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number157480020,157480010,157482500,157480025
compliance-icons compliance-icons
Product Identifier
  • 1-Octanethiol
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Respiratory sensitiser Category 1
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Octanethiol, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.