Learn More
1-Pentanesulfonic acid, sodium salt, 98+%, Thermo Scientific Chemicals
£22.40 - £203.00
Chemical Identifiers
CAS | 22767-49-3 |
---|---|
Molecular Formula | C5H11NaO3S |
Molecular Weight (g/mol) | 174.19 |
MDL Number | MFCD00007541,MFCD00149548 |
InChI Key | ROBLTDOHDSGGDT-UHFFFAOYSA-M |
Synonym | sodium 1-pentanesulfonate, sodium pentane-1-sulfonate, sodium pentanesulfonate, 1-pentanesulfonic acid sodium salt, 1-pentanesulfonic acid, sodium salt, 1-pentylsodiumsulfonate, ipc-alks-5, 1-pentanesulfonate, sodium, pentanesulfonic acid na-salt, pentanesulfonic acidsodiumsalt |
PubChem CID | 23664617 |
IUPAC Name | sodium;pentane-1-sulfonate |
SMILES | [Na+].CCCCCS([O-])(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10473955
|
Thermo Scientific Acros
206110050 |
5 g | Glass bottle |
£22.40
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10688624
|
Thermo Scientific Acros
206110250 |
25 g | Glass bottle |
£67.30
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10698624
|
Thermo Scientific Acros
206111000 |
100 g | Glass bottle |
£203.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
22767-49-3 | |
174.19 | |
ROBLTDOHDSGGDT-UHFFFAOYSA-M | |
23664617 | |
[Na+].CCCCCS([O-])(=O)=O |
C5H11NaO3S | |
MFCD00007541,MFCD00149548 | |
sodium 1-pentanesulfonate, sodium pentane-1-sulfonate, sodium pentanesulfonate, 1-pentanesulfonic acid sodium salt, 1-pentanesulfonic acid, sodium salt, 1-pentylsodiumsulfonate, ipc-alks-5, 1-pentanesulfonate, sodium, pentanesulfonic acid na-salt, pentanesulfonic acidsodiumsalt | |
sodium;pentane-1-sulfonate |
Specifications
22767-49-3 | |
100.0 | |
White to Yellow | |
Authentic | |
C5H11NaO3S | |
MFCD00007541,MFCD00149548 | |
sodium 1-pentanesulfonate, sodium pentane-1-sulfonate, sodium pentanesulfonate, 1-pentanesulfonic acid sodium salt, 1-pentanesulfonic acid, sodium salt, 1-pentylsodiumsulfonate, ipc-alks-5, 1-pentanesulfonate, sodium, pentanesulfonic acid na-salt, pentanesulfonic acidsodiumsalt | |
ROBLTDOHDSGGDT-UHFFFAOYSA-M | |
sodium;pentane-1-sulfonate | |
23664617 | |
≥98% | |
1-Pentanesulfonic acid, sodium salt |
98.5 | |
>300.0°C | |
2 % max. (1 g, 120°C) | |
98+% | |
CH3(CH2)4SO3Na | |
04, III, 23 | |
Solubility in water: soluble. | |
[Na+].CCCCCS([O-])(=O)=O | |
174.19 | |
174.2 | |
Crystals |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
- 1-Pentanesulfonic acid, sodium salt
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only