missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Phenyl-1,2-ethanediol, 97%, Thermo Scientific Chemicals
£20.70 - £156.00
Chemical Identifiers
CAS | 93-56-1 |
---|---|
Molecular Formula | C8H10O2 |
Molecular Weight (g/mol) | 138.17 |
MDL Number | MFCD00003546 |
InChI Key | PWMWNFMRSKOCEY-UHFFFAOYSA-N |
Synonym | 1-phenyl-1,2-ethanediol, styrene glycol, phenylethylene glycol, phenylethanediol, phenyl glycol, 1,2-ethanediol, 1-phenyl, phenyl-1,2-ethanediol, styrolyl alcohol, fenylglycol, 1-phenylethylene glycol |
PubChem CID | 7149 |
IUPAC Name | 1-phenylethane-1,2-diol |
SMILES | C1=CC=C(C=C1)C(CO)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10506941
|
Thermo Scientific Acros
130590050 |
5 g | Glass bottle |
£20.70
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10603332
|
Thermo Scientific Acros
130590250 |
25 g | Plastic bottle |
£61.30
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10550594
|
Thermo Scientific Acros
130591000 |
100 g | Plastic bottle |
£156.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
93-56-1 | |
138.17 | |
PWMWNFMRSKOCEY-UHFFFAOYSA-N | |
7149 | |
C1=CC=C(C=C1)C(CO)O |
C8H10O2 | |
MFCD00003546 | |
1-phenyl-1,2-ethanediol, styrene glycol, phenylethylene glycol, phenylethanediol, phenyl glycol, 1,2-ethanediol, 1-phenyl, phenyl-1,2-ethanediol, styrolyl alcohol, fenylglycol, 1-phenylethylene glycol | |
1-phenylethane-1,2-diol |
Specifications
93-56-1 | |
100.0 | |
White to Beige | |
Authentic | |
C8H10O2 | |
MFCD00003546 | |
15, 8991 | |
PWMWNFMRSKOCEY-UHFFFAOYSA-N | |
1-phenylethane-1,2-diol | |
7149 | |
97% | |
1-Phenyl-1, 2-ethanediol |
96.0 | |
61°C to 67°C | |
272°C to 274°C | |
96% min. (GC) | |
HOCH2CH(C6H5)OH | |
06, 907 | |
1-phenyl-1,2-ethanediol, styrene glycol, phenylethylene glycol, phenylethanediol, phenyl glycol, 1,2-ethanediol, 1-phenyl, phenyl-1,2-ethanediol, styrolyl alcohol, fenylglycol, 1-phenylethylene glycol | |
C1=CC=C(C=C1)C(CO)O | |
138.17 | |
138.17 | |
Crystalline Powder or Flakes |
Safety and Handling
missing translation for 'einecsNumber' : 202-258-1
RUO – Research Use Only