Learn More
1-Trimethylsilylmethanol, 95%, Thermo Scientific Chemicals
£34.27 - £108.54
Chemical Identifiers
CAS | 3219-63-4 |
---|---|
Molecular Formula | C4H12OSi |
Molecular Weight (g/mol) | 104.224 |
MDL Number | MFCD00004596 |
InChI Key | ZQKNBDOVPOZPLY-UHFFFAOYSA-N |
Synonym | trimethylsilyl methanol, 1-trimethylsilylmethanol, hydroxymethyltrimethylsilane, methanol, trimethylsilyl, silane, hydroxymethyl trimethyl, hydroxymethyl trimethylsilane, trimethylhydroxymethylsilane, methanol, 1-trimethylsilyl, trimethylsilyl-methanol, acmc-209hrrShow More |
PubChem CID | 76692 |
IUPAC Name | trimethylsilylmethanol |
SMILES | C[Si](C)(C)CO |
Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
11396856
|
Thermo Scientific Alfa Aesar
L04251.03 |
1 g |
£34.27
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11306866
|
Thermo Scientific Alfa Aesar
L04251.06 |
5 g |
£108.54
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
1-Trimethylsilylmethanol is used as nucleophile to resolve the racemate obtained from enantioselective esterification of naproxen (2-(6-methoxy-2-naphthyl) propionic acid) by lipases. It was also used as a nucleophilic hydroxymethylation agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Trimethylsilylmethanol is used as nucleophile to resolve the racemate obtained from enantioselective esterification of naproxen (2-(6-methoxy-2-naphthyl) propionic acid) by lipases. It was also used as a nucleophilic hydroxymethylation agent.
Solubility
Fully miscible in water.
Notes
Store at 4°C. Incompatible with oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Chemische Identifikatoren
3219-63-4 | |
104.224 | |
ZQKNBDOVPOZPLY-UHFFFAOYSA-N | |
76692 | |
C[Si](C)(C)CO |
C4H12OSi | |
MFCD00004596 | |
trimethylsilyl methanol, 1-trimethylsilylmethanol, hydroxymethyltrimethylsilane, methanol, trimethylsilyl, silane, hydroxymethyl trimethyl, hydroxymethyl trimethylsilane, trimethylhydroxymethylsilane, methanol, 1-trimethylsilyl, trimethylsilyl-methanol, acmc-209hrrShow More | |
trimethylsilylmethanol |
Spezifikation
121°C to 122°C | |
C4H12OSi | |
UN1993 | |
31°C (87°F) | |
Fully miscible in water. | |
C[Si](C)(C)CO | |
104.224 | |
104.23 | |
1.419 | |
1-Trimethylsilylmethanol |
3219-63-4 | |
MFCD00004596 | |
1731486 | |
trimethylsilyl methanol, 1-trimethylsilylmethanol, hydroxymethyltrimethylsilane, methanol, trimethylsilyl, silane, hydroxymethyl trimethyl, hydroxymethyl trimethylsilane, trimethylhydroxymethylsilane, methanol, 1-trimethylsilyl, trimethylsilyl-methanol, acmc-209hrr | |
ZQKNBDOVPOZPLY-UHFFFAOYSA-N | |
trimethylsilylmethanol | |
76692 | |
≥98% | |
0.827 |
Sicherheit und Handhabung
GHS H Statement
H226
Flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
missing translation for 'einecsNumber' : 221-746-5
missing translation for 'rtecsNumber' : PC4100000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.