Learn More
Hamilton™ 10 μL 1700 Series Gas Tight Syringe
The 1700 Series syringes are the Gas Tight version of the 700 series syringes.
Brand:  Hamilton™ CAL80079
 View more versions of this product
Product Code. 17864412
  • £256.00 / Each
Explore more special offers
Description
Description
  • This series of syringes are ideal for dispensing volumes up to 10 μL.
  • A variety of different terminations and needle options are available in this series.
Specifications
Specifications
Autoclavable
22s G
0.006 in.
0.0283 in.
0.460 mm
0.72 mm
239°F
50°F
10°C
10 μL
Blunt
Stainless Steel
0.018 in.
0.15 mm
0.260 in.
6.60 mm
2 in.
115°C
1701 Syringe
1
51 mm
Removable