Learn More
Hamilton™ 10 μL 1700 Series Gas Tight Syringe
The 1700 Series syringes are the Gas Tight version of the 700 series syringes.
£234.26 - £265.00
Specifications
Diameter (English) Inner | 0.018 in. |
---|---|
Diameter (English) Outer | 0.260 in. |
Diameter (Metric) Inner | 0.460 mm |
Diameter (Metric) Outer | 6.60 mm |
Min. Temperature (English) | 50°F |
Product Code | Brand | Needle Gauge | Needle Point Style | Connection Type | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Needle Gauge | Needle Point Style | Connection Type | Price | Quantity & Availability | |||
17864252
|
Hamilton™
CAL80088 |
23s G | Custom | Cemented |
£262.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17894322
|
Hamilton™
CAL80308 |
Custom | Custom | Cemented |
£252.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17804362
|
Hamilton™
CAL80030 |
26S (No Needle Included) | 2: Curved, Beveled (No Needle Included) | Removable |
£241.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17814362
|
Hamilton™
CAL80011 |
26s G | Sharp, Bevel, Curved, Non-coring | Cemented |
£265.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17844412
|
Hamilton™
CAL80080 |
26s G | Blunt | Cemented |
£253.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17834412
|
Hamilton™
CAL80075 |
26s G | Cone | Cemented |
£234.26
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17884242
|
Hamilton™
CAL80039 |
No Needle Included | No Needle Included | Removable |
£257.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17864412
|
Hamilton™
CAL80079 |
22s G | Blunt | Removable |
£256.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17854252
|
Hamilton™
CAL80087 |
23s G | Cone | Removable |
£262.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17864242
|
Hamilton™
CAL80065 |
26s G | Sharp, Bevel, Curved, Non-coring | Removable |
£245.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- This series of syringes are ideal for dispensing volumes up to 10 μL.
- A variety of different terminations and needle options are available in this series.
Specifications
0.018 in. | |
0.460 mm | |
50°F | |
1 |
0.260 in. | |
6.60 mm | |
10°C | |
10 μL |