Learn More
Wheaton™ 10 μL Inoculation Loop
Inoculation Loop for delivering a known volume of fluid sample to enable accurate microbiological analysis
Brand: Wheaton™ 711420
37.97 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- For dispersing samples on agar surface
- Meet quantitative standards for management of microbiological samples
Specifications
Single-use | |
ABS | |
Standard | |
500 | |
Not Autoclavable |
10 μL | |
Sterile | |
Blue | |
Packaged in peel pouch |
Product Content Correction
Product Title
Wheaton™ 10 μL Inoculation Loop > Size: Standard
Spot an opportunity for improvement?Share a Content Correction