Learn More
BRAND™ Inoculation Loop with Needle
Inoculation Loop with Needle designed for inoculation of nutrient media.
Brand: BRAND™ 452215
This item is not returnable.
View return policy
Description
- For inoculation of nutrient media
- Available with loop at one end and needle at the other
- High flexibility permits gentle streaking without damaging the nutrient surface
- γ-ray sterilized
Specifications
1 μL | |
Gamma Sterilized | |
Needle |
Polystyrene | |
Yellow | |
For inoculation of nutrient media |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Product Title
BRAND™ Inoculation Loop with Needle > Sterility: Sterile
Spot an opportunity for improvement?Share a Content Correction