Learn More
Brand™ Polystyrene Inoculation Loop
Permits gentle streaking without damaging the nutrient surface. BRAND™ Polystyrene Inoculation Loops are highly flexible, with loop at one end and needle at the other or loops at both ends.
Brand: Brand™ 452201
55.64 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Description
- Manufactured from polystyrene.
- For inoculation of nutrient media.
- Available either with loop at one end and needle at the other or loops at both ends.
- High flexibility permits gentle streaking without damaging the nutrient surface.
- y-ray sterilized.
Specifications
Single-use | |
Polystyrene | |
White | |
Needle |
1 μL | |
Sterile | |
Inoculating Loop | |
1000/Pk. |
Special Offers
-
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24155Expires: 2025-04-30
Days
43Hours
23Minutes
13Seconds
2